Merck
  • Solution NMR of polypeptides hyperpolarized by dynamic nuclear polarization.
Solution NMR of polypeptides hyperpolarized by dynamic nuclear polarization.
Analytical chemistry (2011-06-10)
Mukundan Ragavan, Hsueh-Ying Chen, Giridhar Sekar, Christian Hilty
ABSTRACT
Hyperpolarization of nuclear spins through techniques such as dynamic nuclear polarization (DNP) can greatly increase the signal-to-noise ratio in NMR measurements, thus eliminating the need for signal averaging. This enables the study of many dynamic processes which would otherwise not be amenable to study by NMR spectroscopy. A report of solid- to liquid-state DNP of a short peptide, bacitracin A, as well as of a full-length protein, L23, is presented here. The polypeptides are hyperpolarized at low temperature and dissolved for NMR signal acquisition in the liquid state in mixtures of organic solvent and water. Signal enhancements of 300-2000 are obtained in partially deuterated polypeptide when hyperpolarized on (13)C and of 30-180 when hyperpolarized on (1)H. A simulated spectrum is used to identify different resonances in the hyperpolarized (13)C spectra, and the relation between observed signal enhancement for various groups in the protein and relaxation parameters measured from the hyperpolarized samples is discussed. Thus far, solid- to liquid-state DNP has been used in conjunction with small molecules. The results presented here, however, demonstrate the feasibility of hyperpolarizing larger proteins, with potential applications toward the study of protein folding or macromolecular interactions.
Product Description
Supelco
Bacitracin A, VETRANAL®, analytical standard, main stereoisomer ~50 %