Skip to Content
Merck
All Photos(1)

Key Documents

16-157

Sigma-Aldrich

Protein A Agarose/Salmon Sperm DNA, 2.5 mL

for use in chromatin immunoprecipitations (ChIP assays)

Synonym(s):

ChIP agarose beads, ChIP agaraose A beads, ChIP assays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

Staphylococcus aureus

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable

suitability

suitable for immunoprecipitation

shipped in

wet ice

General description

Protein A covalently bound to agarose by alkylamine linkage containing sonicated Salmon Sperm DNA. The salmon sperm DNA blocks non-specific interactions with the beads for use in chromatin IP (ChIP) assays. Note part number 16-157C present in the ChIP kits is not available separately, please purchase 16-157.
Protein A from Staphylococcus aureus has a very high affinity for the Fc regions of IgG molecules. Immoblisation of protein A on beads creates an affinity resin which can be used to purify IgG fractions from crude serum, ascites fluid or cell cuture media.

Application

Suitable for use in chromatin immunoprecipitation (ChIPs) assays

Quality

Routinely evaluated in a Chromatin Immunoprecipitation by pulling down DNA cross-linked to acetylated histones; subsequent detection was performed using anti-Acetyl Histone H4, ChIPs Grade (06-866)

Physical form

10mM Tris-HCl, 1mM EDTA, pH 8.0 containing 0.05% sodium azide.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Generation of p53 target database via integration of microarray and global p53 DNA-binding site analysis.
Suxing Liu, Asra Mirza, Luquan Wang
Methods in Molecular Biology null
Mohammad Kamran et al.
Journal of immunology (Baltimore, Md. : 1950), 205(12), 3311-3318 (2020-11-15)
IL-13 plays a critical role in mediating many biological processes responsible for allergic inflammation. Mast cells express Il13 mRNA and produce IL-13 protein in response to antigenic stimulation. Enhancers are essential in promoting gene transcription and are thought to activate
A note on penicillin resistance and beta-lactamase production in strains of Staphylococcus aureus recovered from different clinical materials in Nsukka, Nigeria.
Oguike, J U, et al.
The Journal of Communicable Diseases, 23, 200-201 (1991)
Requirement of Smad3 and CREB-1 in mediating transforming growth factor-beta (TGF beta) induction of TGF beta 3 secretion.
Guangming Liu, Wei Ding, Jill Neiman, Kathleen M Mulder
The Journal of Biological Chemistry null
Guodong Huang et al.
Autophagy, 12(8), 1292-1309 (2016-05-14)
Autophagy is a highly conserved intracellular degradation system, and recently was shown to display circadian rhythms in mice. The mechanisms underlying circadian regulation of autophagy, however, are still unclear. Here, we observed that numbers of autophagosomes and autolysosomes exhibit daily

Related Content

Protein and nucleic acid interaction reagents and resources for investing protein-RNA, protein-DNA, and protein-protein interactions and associated applications.

Protein and nucleic acid interaction reagents and resources for investing protein-RNA, protein-DNA, and protein-protein interactions and associated applications.

Protein and nucleic acid interaction reagents and resources for investing protein-RNA, protein-DNA, and protein-protein interactions and associated applications.

Protein and nucleic acid interaction reagents and resources for investing protein-RNA, protein-DNA, and protein-protein interactions and associated applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service