Skip to Content
Merck

16-201

Protein G Agarose/Salmon Sperm DNA, 2.5 mL

for use in chromatin immunoprecipitations (ChIP assays)

Synonym(s):

ChIP agarose beads, ChIP agaraose G beads

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
eCl@ss:
32160405
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable

suitability

suitable for immunoprecipitation

shipped in

wet ice

General description

Recombinant Protein G covalently bound to agarose by cyanogen bromide linkage. Sonicated salmon sperm DNA blocks non-specific DNA binding sites on protein G agarose when collecting immunocomplexes for chromatin immunoprecipitations (ChIP assays).
Recombinant protein G has a very high affinity for the Fc regions of IgG molecules. Immoblisation of protein G on beads creates an affinity resin which can be used to purify IgG fractions from crude serum, ascites fluid or cell cuture media.

Packaging

2.5 mL packed beads provided as a 50% gel slurry for a final volume of 5 mL.

Physical form

2.5 mL packed beads containing 1 mg sonicated salmon sperm DNA, 2.5 mg BSA and approximately 5 mg recombinant Protein G. Provided as a 50% gel slurry for a final volume of 5 mL per vial. Suspended in 10 mM Tris-HCl, 1 mM EDTA, pH 8.0, containing 0.05% sodium azide. Liquid suspension.

Preparation Note

1 year at 4°C from date of shipment.

Analysis Note

Routinely evaluated in Chromatin Immunoprecipitation for use to pre-clear the supernatant from sodium butyrate treated HeLa cells and subsequent immunoprecipitation of DNA cross-linked to acetylated histones

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

10 - Combustible liquids

wgk

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyster K Nanan et al.
Nucleic acids research, 45(22), 12780-12797 (2017-12-16)
Actively transcribed genes adopt a unique chromatin environment with characteristic patterns of enrichment. Within gene bodies, H3K36me3 and cytosine DNA methylation are elevated at exons of spliced genes and have been implicated in the regulation of pre-mRNA splicing. H3K36me3 is
Fen Hu et al.
Experimental and therapeutic medicine, 15(1), 569-575 (2018-02-06)
Human embryo lung cellular protein interacting with severe acute respiratory syndrome-coronavirus nonstructural protein-10 (HEPIS) is a novel transcriptional repressor, the expression profile and promoter activity of which have not been well studied. In the present study, in situ hybridization of
Hidenori Kaneoka et al.
Cytotechnology, 57(2), 123-128 (2008-11-13)
The glucocorticoid receptor regulates liver-specific expression of the tryptophan oxygenase gene through glucocorticoid responsive elements located -0.45 and -1.2 kb from the transcription start site. However, the hormone-mediated induction is restricted to adult hepatocytes, and fetal hepatocytes are unable to
Yang Tang et al.
Molecular plant, 10(2), 260-273 (2016-11-24)
Light is a crucial environmental signal that promotes photomorphogenesis, the developmental process with a series of light-dependent alterations for plants to adapt various external challenges. Chromatin modification has been proposed to be involved in such light-mediated growth, but the underlying
Kyung-Soon Park et al.
Stem cells (Dayton, Ohio), 31(2), 282-292 (2012-11-22)
Self-renewal and pluripotency are hallmark properties of pluripotent stem cells, including embryonic stem cells (ESCs) and iPS cells. Previous studies revealed the ESC-specific core transcription circuitry and showed that these core factors (e.g., Oct3/4, Sox2, and Nanog) regulate not only

Related Content

"Epigenetics describes heritable changes in gene expression caused by non-genetic mechanisms instead of by alterations in DNA sequence. These changes can be cell- or tissue-specific, and can be passed on to multiple generations. Epigenetic regulation enriches DNAbased information, allowing a cell to vary its response across diverse biological and environmental contexts. Although epigenetic mechanisms are primarily centered in the nucleus, these mechanisms can be induced by environmental signals such as hormones, nutrients, stress, and cellular damage, pointing to the involvement of cytoplasmic and extracellular factors in epigenetic regulation."

Cancer is a complex disease manifestation. At its core, it remains a disease of abnormal cellular proliferation and inappropriate gene expression. In the early days, carcinogenesis was viewed simply as resulting from a collection of genetic mutations that altered the gene expression of key oncogenic genes or tumor suppressor genes leading to uncontrolled growth and disease (Virani, S et al 2012). Today, however, research is showing that carcinogenesis results from the successive accumulation of heritable genetic and epigenetic changes. Moreover, the success in how we predict, treat and overcome cancer will likely involve not only understanding the consequences of direct genetic changes that can cause cancer, but also how the epigenetic and environmental changes cause cancer (Johnson C et al 2015; Waldmann T et al 2013). Epigenetics is the study of heritable gene expression as it relates to changes in DNA structure that are not tied to changes in DNA sequence but, instead, are tied to how the nucleic acid material is read or processed via the myriad of protein-protein, protein-nucleic acid, and nucleic acid-nucleic acid interactions that ultimately manifest themselves into a specific expression phenotype (Ngai SC et al 2012, Johnson C et al 2015). This review will discuss some of the principal aspects of epigenetic research and how they relate to our current understanding of carcinogenesis. Because epigenetics affects phenotype and changes in epigenetics are thought to be key to environmental adaptability and thus may in fact be reversed or manipulated, understanding the integration of experimental and epidemiologic science surrounding cancer and its many manifestations should lead to more effective cancer prognostics as well as treatments (Virani S et al 2012).

Global Trade Item Number

SKUGTIN
16-20104053252326370

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service