Skip to Content
Merck
All Photos(6)

Documents

05-1242-S

Sigma-Aldrich

Anti-trimethyl-Histone H3 (Lys9) Antibody, clone 6F12-H4, Trial Size

clone 6F12-H4, from mouse

Synonym(s):

H3K9me3, Histone H3 (trimethyl K9), H3 histone family, member T, Histone 3, H3, Histone cluster 3, H3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

6F12-H4, monoclonal

species reactivity

mouse, human

technique(s)

ChIP: suitable (ChIP-seq)
dot blot: suitable
immunofluorescence: suitable
inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys9)

Gene Information

human ... H3C1(8350)
mouse ... H3C1(360198)

Related Categories

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped around the octamer, forming a nucleosome, the basic subunit of chromatin. Histone modifications regulate DNA transcription, repair, recombination, and replication. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. These modifications can alter local chromatin architecture, or recruit trans-acting factors that recognize specific histone modifications (the "histone code" hypothesis). Trimethylation of histone H3 on Lys9 (H3K9me3) is one of the most highly studied epigenetic marks. H3K9me3 functions in the repression of euchromatic genes, and in epigenetic control of heterochromatin assembly, most likely via acting as a recognition motif for the binding of chromatin-associated proteins, such as Swi6 or HP1α/β. The enzymes responsible for H3K9me3 formation are SUV39H1 and SUV39H2.

Specificity

Based on sequence homology, broad species cross-reactivity is expected with all mammals, Drosophila, Xenopus, and Arabidopsis.
Recognizes trimethyl-histone H3 (Lys9), Mr 17 kDa. Cross-reacts with dimethyl histone H3(Lys9) at higher concentrations.

Immunogen

Epitope: Trimethyl Lys9
KLH-conjugated, branched synthetic peptide containing the sequence (QTARme3K- STGGKA)2-KC, in which me3K corresponds to trimethyl lysine 9 of human histone H3. Data suggests this antibody may require the presence of the glutamine residue in position 5 for binding.

Application

"Chromatin Immunoprecipitation (ChIP):
Representative data from a previous lot. Sonicated 3T3 L1 chromatin was subjected to chromatin immunoprecipitation using anti- trimethyl-histone H3 (Lys9) and the Magna ChIP G (Cat. #17-611) Kit. Successful immunoprecipitation of trimethylhistone H3 (Lys9) associated DNA fragments was verified by qPCR using primers flanking the p16 promoter.

Peptide Inhibition Analysis:
Peptide blocking assay demonstrates distinct preference of the antibody for the trimethyl form vs. the dimethyl form.


Chromatin Immunoprecipitation (ChIP):
ChIP analysis of known chromosomal Suv39h targets (H3K9me3 in major satellites, mouseES cells).


Dot Blot Analysis:
Dot-blot analysis demonstrating specificity of anti-H3K9me3, clone 6F12-H4 for trimethyl Lys9 of histone H3.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-trimethyl-Histone H3 (Lys9) Antibody, clone 6F12-H4 (mouse monoclonal antibody), Trial Size / Pack validated in ChIP, PIA, IF, DB, ChIP-seq, WB to detect trimethyl-Histone H3 (Lys9) also known as H3K9me3, Histone H3 (tri methyl K9).

Quality

Routinely evaluated by Western Blot on HeLa acid extracts.

Western Blot Analysis: A 0.5 – 5 μg dilution of this lot detected trimethyl histone H3 (Lys9) in HeLa acid extracts.

Target description

~17 kDa

Physical form

Format: Purified
Purified
Purified antibody supplied at 1 mg/ml in 0.1 M Tris-glycine, pH 7.4, 150 mM NaCl, 0.05% NaN3.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
For maximum recovery of product, centrifuge the vial prior to removing the cap. Avoid freezing, which can damage IgG and affect product performance.

Analysis Note

Control
HeLa Acid Extract

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Chiang et al.
Cell reports, 28(12), 3212-3223 (2019-09-19)
Lamina-associated domains (LADs) cover a large part of the human genome and are thought to play a major role in shaping the nuclear architectural landscape. Here, we perform polymer simulations, microscopy, and mass spectrometry to dissect the roles played by
Viviana Barra et al.
Genes, 13(7) (2022-07-28)
Background: Induced senescence could be exploited to selectively counteract the proliferation of cancer cells and target them for senolysis. We examined the cellular senescence induced by curcumin and whether it could be targeted by fisetin and quercetin, flavonoids with senolytic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service