Skip to Content
Merck
All Photos(2)

Documents

SAB4200808

Sigma-Aldrich

Anti-Myosin Light Chain Kinase antibody, Mouse monoclonal

clone K36, purified from hybridoma cell culture

Synonym(s):

Kinase-related protein (KRP), MLCK, Myosin light chain kinase, Smooth muscle, Telokin, smMLCK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

K36, monoclonal

form

buffered aqueous solution

mol wt

~160 kDa

species reactivity

bovine, human, mouse, chicken, pig, turkey

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoblotting: 0.25-0.5 μg/mL using chicken gizzard extract

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Myosin Light Chain Kinase (MLCK) also known as MYLK is a Ca2+/calmodulin dependent myosin light chain phosphorylating agent. This enzyme plays a major role in the phosphorylation of the regulatory light chains of myosin which are essential for the shortening and tension development of smooth muscle cells resulting in smooth muscle contraction. Myosin light chain kinase are widely expressed in many different tissues and cells of eukaryote species. There are two genes mylk1 and mylk2 encoding the MLCK protein, mylk2 is exclusively expressed in skeletal muscle cells.

Immunogen

Purified chicken gizzard myosin light chain kinase.

Application

Monoclonal Anti-Myosin Light Chain Kinase recognizes the myosin light chain kinase of smooth muscle and from non-muscle cells such as cultured fibroblasts. Reactivity has been observed with myosin light chain kinase from chicken, turkey, bovine, human, mouseand pig origin. The antibody is recommended to use in various immunological techniques, including Immunoblotting (~160kDa), Immunohistochemistry and immunofluorescence.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Other Notes

This product is for R&D use only, not for drug, household, or other uses.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

So Youn Moon et al.
Molecular biology of the cell, 34(9), ar89-ar89 (2023-06-21)
Matrix stiffening is ubiquitous in solid tumors and can direct epithelial-mesenchymal transition (EMT) and cancer cell migration. Stiffened niche can even cause poorly invasive oral squamous cell carcinoma (OSCC) cell lines to acquire a less adherent, more migratory phenotype, but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service