Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1996

Sigma-Aldrich

Anti-ACLY Antibody, clone 1B16 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

ATP-citrate synthase;EC:2.3.3.8;ATP-citrate (pro-S-)-lyase;ACL;Citrate cleavage enzyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B16, recombinant monoclonal

description

1B16 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 120.84 kDa

purified by

using Protein A

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ACLY(47)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B16 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects ATP citrate synthase (ACLY). It targets an epitope within 20 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of human ATP-citrate synthase (ACLY).

Application

Quality Control Testing

Evaluated by Western Blotting in NIH3T3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ACLY in NIH3T3 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected ACLY in lysates from Raji cells and Rat brain tissue.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected ACLY in human pancreas tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected ACLY in U2OS cells.

Affinity Binding Assay: A representative lot of this antibody bound ATP Citrate Synthase (ACLY) peptide with a KD of 5.5 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

ATP-citrate synthase (UniProt: P53396; also known as EC:2.3.3.8, ATP-citrate (pro-S-)-lyase, ACL, Citrate cleavage enzyme ACLY) is encoded by the ACLY gene (Gene ID: 47) in human. ACLY is a homotetrameric enzyme that catalyzes the conversion of mitochondrial citrate into oxaloacetate and acetyl-CoA. This step is a major source of cytosolic acetyl-CoA that is used in the biosynthetic pathways of fatty acids, cholesterol, and acetylcholine. ACLY is strongly expressed in liver, adipose tissue, and cholinergic neurons. Its activity is stimulated by glucose 6-phosphate, fructose 6-phosphate, fructose 2,6-bisphosphate, and fructose 1,6-bisphosphate. ACLY is phosphorylated by PKA and GSK3 in a sequential manner, which results in its activation. Phosphorylation on threonine 447 and serine 451 by GSK3 is reported to depend on the phosphorylation of serine 455 by PKA. ACLY also undergoes acetylation at lysine 540, 546, and 554 by lysine aetyltransferase 2B ( KAT2B/PCAF). These acetylations are promoted by glucose and lead to its stabilization, probably by inhibiting ubiquitination at these sites. Overexpression of ACLY has been reported many lipogenic tumors, including prostate, liver, breast, and ovarian cancers. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Verschueren, KHG., et al. (2019). Nature. 568; 571-575; Lin, R. et al. (2013). Mol Cell. 51(4); 506-518; Potapova, IA., et al. (2000). Biochemistry. 39(5); 1169-1179).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service