Skip to Content
Merck
All Photos(1)

Documents

04-1462

Sigma-Aldrich

Anti-hnRNP F Antibody, clone 3H4

clone 3H4, from mouse

Synonym(s):

heterogeneous nuclear ribonucleoprotein F, nucleolin-like protein, HnRNP F protein, Nucleolin-like protein mcs94-1, mcs94-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3H4, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HNRNPF(3185)

Related Categories

General description

The hnRNP F protein is member of a subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs).
The hnRNPs are RNA binding proteins that complex with heterogeneous nuclear RNA (hnRNA) and associate with pre-mRNAs in the nucleus. These complexes are associated with pre-mRNA processing and other aspects of mRNA metabolism and transport. There are more than 20 different heterogeneous nuclear ribonucleoproteins (hnRNPs) associated with pre-mRNAs in the nucleus of mammalian cells. These different hnRNPs display a range of affinities for a variety of ribonucleotidehomopolymers and ssDNA sequences. In constrast, hnRNP F protein as well as hnRNP H (a highly related protein) appear to bind only to poly(rG). These unique RNA-binding properties, as well as the amino acid sequence and other data showing a distinct intranuclear localization of hnRNP F and hnRNP H suggest they are unique from other hnRNP and play an important role in the processing of RNAs containing guanosine-rich sequences.

Specificity

This antibody recognizes hnRNP F.

Immunogen

Epitope: Unknown
Recombinant protein corresponding to human hnRNP F.

Application

Immunoprecipitation Analysis:
A representative lot was used by an independent laboratory for IP. (Matunis, M.J., et al. (1994). Nucleic Acids Res. 22(6):1059–1067.)

Immunofluorescence Analysis:
A representative lot was used by an independent laboratory in IF. (Matunis, M.J., et al. (1994). Nucleic Acids Res. 22(6):1059–1067.)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Use Anti-hnRNP F Antibody, clone 3H4 (Mouse Monoclonal Antibody) validated in WB, IP, ICC to detect hnRNP F also known as heterogeneous nuclear ribonucleoprotein F.

Quality

Evaluated by Western Blot in K562 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected hnRNP F on 10 µg of K562 cell lysate.

Target description

Calculated: 46 kDa Observed: ~53 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
K562 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Miao et al.
Science advances, 8(51), eabq7289-eabq7289 (2022-12-24)
Understanding how long noncoding RNAs (lncRNAs) cooperate with splicing factors (SFs) in alternative splicing (AS) control is fundamental to human biology and disease. We show that metastasis-associated lung adenocarcinoma transcript 1 (MALAT1), a well-documented AS-implicated lncRNA, regulates AS via two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service