Skip to Content
Merck
All Photos(1)

Key Documents

A-913

Supelco

7-Aminonitrazepam solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O
CAS Number:
Molecular Weight:
251.28
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

O=C1NC2=CC=C(N)C=C2C(C3=CC=CC=C3)=NC1

InChI

1S/C15H13N3O/c16-11-6-7-13-12(8-11)15(17-9-14(19)18-13)10-4-2-1-3-5-10/h1-8H,9,16H2,(H,18,19)

InChI key

OYOUQHVDCKOOAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

7-Aminonitrazepam is a major urinary metabolite of nitrazepam, a benzodiazepine used to treat insomnia and sold under the trade names Mogadon and Alodorm. This certified solution standard is suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug monitoring of nitrazepam use.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic determination of nitrazepam and its metabolites in human urine.
T Kozu
Journal of chromatography, 310(1), 213-218 (1984-09-14)
M Tomita et al.
Journal of chromatography, 621(2), 249-255 (1993-11-24)
We applied micellar electrokinetic capillary chromatography to simultaneous separation and determination of nitrazepam and its major metabolites, 7-aminonitrazepam and 7-acetamidonitrazepam, in spiked urine. Prior to electrophoresis, the three compounds were successfully extracted from the spiked urine with commercial disposable solid-phase
Fumio Moriya et al.
Forensic science international, 131(2-3), 108-112 (2003-02-19)
We report a case of nitrazepam poisoning in which the distribution of nitrazepam and 7-aminonitrazepam was determined in body fluids and tissues. A 52-year-old woman was found dead in a shallow ditch (approximately 5 cm in depth), in the winter.
B J Hart et al.
Methods and findings in experimental and clinical pharmacology, 10(1), 21-26 (1988-01-01)
The stability of some selected benzodiazepines in saliva has been studied. The benzodiazepines nitrazepam and clonazepam were found to be unstable in saliva at room temperature and nitrazepam was converted into 7-aminonitrazepam. The conversion rate of nitrazepam was strongly dependent
Yu Zhu et al.
Se pu = Chinese journal of chromatography, 20(5), 394-397 (2005-12-20)
A highly sensitive method has been developed for the analysis of 7-aminonitrazepam (7-ANIZ), the major metabolite of nitrazepam, in urine by trimethylsilyl derivatization-gas chromatography/mass spectrometry. Urine samples were extracted with ethyl ether-ethyl acetate (99:1, volume ratio). The extracts were derivatized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service