Skip to Content
Merck

Skip To

B-033

Benzodiazepine Multi-component Mixture-8 solution

250 μg/mL each component in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116107
EC Number:
200-835-2
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

Alprazolam , Clonazepam , Diazepam , Flunitrazepam , Lorazepam , Nitrazepam , Oxazepam , Temazepam

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIC/IV (Portugal), kontrollierte Droge in Deutschland

concentration

250 μg/mL each component in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

multi-component solution

storage temp.

−20°C

General description

A Certified Spiking Solution® of routinely tested benzodiazepines alprazolam, clonazepam, diazepam, flunitrazepam, lorazepam, nitrazepam, oxazepam and temazepam. This multi-component mix is suitable for use in preparation of linearity standards, calibrators, or controls as well as for other critical quantitative applications such as internal proficiency testing or for OEM use in preparation of reagents, assay development, and instrument IQ/OQ. Certified concentration values for each component provides increased efficiency from less time remaking pools due to dilution errors or rerunning samples.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C-111C-109A-144
application(s)

forensics and toxicology

application(s)

clinical testing
forensics and toxicology

application(s)

clinical testing
forensics and toxicology

application(s)

clinical testing

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

form

liquid

form

liquid

form

liquid

form

liquid

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

storage temp.

−20°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

packaging

ampule of 1 mL

packaging

ampule of 1 mL

packaging

ampule of 1 mL

packaging

ampule of 1 mL


Analyte

Description

  • Alprazolam

  • Clonazepam

  • Diazepam

  • Flunitrazepam

  • (±)-Lorazepam

  • Nitrazepam

  • Oxazepam

  • Temazepam

See All (8)

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters