Skip to Content
Merck

Skip To

B-905

Benzodiazepine Internal Standard-3 solution

0.5-1.0 mg/mL in acetonitrile, ampule of 1.0 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116107
EC Number:
200-835-2
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

7-Aminoclonazepam-D4, 0.5 mg/mL , 7-Aminoflunitrazepam-D7, 0.5 mg/mL , α-Hydroxytriazolam-D4, 1.0 mg/mL

packaging

ampule of 1.0 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland)

concentration

0.5-1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

General description

A stable-labeled internal multi-component solution mix of routinely tested benzodiazepines 7-aminoclonazepam, 7-aminoflunitrazepam, and α-hydroxytriazolam. This internal standard mix is suitable for quantitation of benzodiazepine levels in urine, serum or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Analyte

Description

  • 7-Aminoclonazepam-d4 .5 %

  • 7-Aminoflunitrazepam-d7 .5 mg/mL

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters