Skip to Content
Merck
All Photos(1)

Key Documents

A-912

Supelco

7-Aminoflunitrazepam solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14FN3O
CAS Number:
Molecular Weight:
283.30
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

FC1=CC=CC=C1C2=NCC(N(C)C3=CC=C(N)C=C23)=O

InChI

1S/C16H14FN3O/c1-20-14-7-6-10(18)8-12(14)16(19-9-15(20)21)11-4-2-3-5-13(11)17/h2-8H,9,18H2,1H3

InChI key

LTCDLGUFORGHGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

As a major metabolite of flunitrazepam, 7-aminoflunitrazepam is a biomarker for use of the parent drug. Flunitrazepam is an intermediate-acting benzodiazepine, typically used in the treatment of severe insomnia. This certified solution standard is suitable for numerous GC/MS or LC/MS testing applications in forensic analysis, clinical toxicology, urine drug testing, or pain prescription monitoring.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffery Hackett et al.
Forensic science international, 157(2-3), 156-162 (2005-06-16)
The forensic toxicology community has recognized flunitrazepam and its metabolite (7-aminoflunitrazepam) as compounds of concern for several years. In this procedure, the analytes were extracted from whole blood and urine onto single mode solid phase cartridges (butyl) using nitrazepam as
H L Lord et al.
Journal of pharmaceutical and biomedical analysis, 40(3), 769-780 (2005-12-15)
We report on the development of solid phase microextraction probes for drug analysis, prepared with antibodies specific for benzodiazepines covalently immobilized to the surface. In the technique, immobilized antibody probes are exposed to a sample containing the drug for 30
Chiung-Wei Huang et al.
Journal of chromatography. A, 1110(1-2), 240-244 (2006-02-18)
A sweeping technique, in conjunction with micellar electrokinetic chromatography, for the simultaneous determination of flunitrazepam and its major metabolites, 7-aminoflunitrazepam and N-desmethylflunitrazepam, is described. The optimized conditions for the sweeping and separation were a pH 9.5 buffer, 25mM borate, 50mM
I Gafni et al.
Journal of clinical psychopharmacology, 23(2), 169-175 (2003-03-18)
Flunitrazepam, a hypnotic benzodiazepine, is widely prescribed around the world for the treatment of insomnia and as a preanesthetic. In vitro studies have shown that the metabolism of flunitrazepam to desmethylflunitrazepam and 3-hydroxyflunitrazepam is mediated in part by the polymorphic
Masaru Terada et al.
Legal medicine (Tokyo, Japan), 5 Suppl 1, S96-S100 (2003-08-26)
A method for the determination of flunitrazepam (FNZ) and 7-aminoflunitrazepam (7-AFNZ) in human serum was developed with ion trap gas chromatography (GC)-tandem mass spectrometry. The 7-AFNZ was derivatizated with 50 microl trifluoroacetic anhydride (TFAA), 60 degrees C-20 min. EI mass

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service