Skip to Content
Merck
All Photos(1)

Documents

W279706

Sigma-Aldrich

Octanal

≥95%, FCC, FG

Synonym(s):

Aldehyde C8, Caprylic aldehyde, Octyl aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CHO
CAS Number:
Molecular Weight:
128.21
FEMA Number:
2797
Beilstein:
1744086
EC Number:
Council of Europe no.:
97
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.009
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

vapor pressure

2 mmHg ( 20 °C)
850 mmHg ( 25 °C)
850 mmHg ( 25 °C)

Assay

≥95%

refractive index

n20/D 1.421 (lit.)

bp

171 °C (lit.)

mp

12-15 °C (lit.)

density

0.82 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fatty; green; orange; waxy; citrus

SMILES string

[H]C(=O)CCCCCCC

InChI

1S/C8H16O/c1-2-3-4-5-6-7-8-9/h8H,2-7H2,1H3

InChI key

NUJGJRNETVAIRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minh Q Nguyen et al.
PloS one, 7(7), e41899-e41899 (2012-08-01)
In mammals, odorants are detected by a large family of receptors that are each expressed in just a small subset of olfactory sensory neurons (OSNs). Here we describe a strain of transgenic mice engineered to express an octanal receptor in
R C Araneda et al.
Nature neuroscience, 3(12), 1248-1255 (2000-12-02)
An odor perception is the brain's interpretation of the activation pattern of many peripheral sensory neurons that are differentially sensitive to a wide variety of odors. The sensitivity of these neurons is determined by which of the thousand or so
Yong Jie Li et al.
Environmental science & technology, 42(19), 7138-7145 (2008-10-23)
Atmospheric accretion reactions of octanal with sulfuric acid as a catalyst were investigated in bulk liquid-liquid experiments and gas-particle experiments. In bulk studies, trioxane, alpha,beta-unsaturated aldehyde, and trialkyl benzene were identified by gas chromatography-mass spectrometry as major reaction products with
Ricardo C Araneda et al.
The Journal of physiology, 555(Pt 3), 743-756 (2004-01-16)
Several lines of evidence suggest that odorants are recognized through a combinatorial process in the olfactory system; a single odorant is recognized by multiple receptors and multiple odorants are recognized by the same receptor. However few details of how this
Mariel A Kerr et al.
Nature neuroscience, 9(4), 484-486 (2006-03-21)
Primary olfactory sensory neurons (OSNs) impart both molecular and functional organization to the olfactory bulb. Because OSNs can be selectively activated by odorants in vivo, we sought to determine whether odorant experience alters the cellular dynamics of specific OSNs or

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service