Skip to Content
Merck
All Photos(1)

Key Documents

W280003

Sigma-Aldrich

1-Octanol

≥98%, FCC, FG

Synonym(s):

Alcohol C8, Capryl alcohol, Octyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OH
CAS Number:
Molecular Weight:
130.23
FEMA Number:
2800
Beilstein:
1697461
EC Number:
Council of Europe no.:
54
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.006
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

vapor density

4.5 (vs air)

vapor pressure

0.14 mmHg ( 25 °C)

Assay

≥98%

form

liquid

autoignition temp.

523 °F

refractive index

n20/D 1.429 (lit.)

bp

196 °C (lit.)

mp

−15 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fatty; green; citrus; waxy; floral; sweet

SMILES string

CCCCCCCCO

InChI

1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3

InChI key

KBPLFHHGFOOTCA-UHFFFAOYSA-N

Gene Information

mouse ... Chrna1(11435)

Looking for similar products? Visit Product Comparison Guide

General description

1-Octanol is one of the key flavor constituents of mushrooms.

Application

  • Noncontact In Situ Multidiagnostic NMR/Dielectric Spectroscopy: Research highlighted the application of 1-octanol in enhancing spectroscopic analysis techniques, providing noncontact, in situ measurements for complex biological samples (Morais AF, Radhakrishnan S, Arbiv G, et al., 2024) Anal Chem.

  • Development of a dispersive liquid-liquid microextraction method for the determination of plastic additives in seawater: Demonstrates the role of 1-octanol in environmental monitoring through the development of a microextraction method for assessing plastic additives in marine environments (González-Castro MJ, Uribe-Ares J, Muniategui-Lorenzo S, et al., 2024) Anal Methods.

  • Application of novel metal organic framework-deep eutectic solvent/molecularly imprinted polymer multiple monolithic fiber for solid phase microextraction of amphetamines and modafinil in unauthorized medicinal supplements with GC-MS: This study utilizes 1-octanol in a novel solid phase microextraction method, highlighting its utility in forensic and pharmaceutical analysis (Kardani F, Khezeli T, Shariati S, et al., 2024) J Pharm Biomed Anal.

Biochem/physiol Actions

Blocks T-type Ca2+ channels

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

187.7 °F

Flash Point(C)

86.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavor compounds in king oyster mushrooms Pleurotus eryngii.
Mau JL, et al.
Journal of Agricultural and Food Chemistry, 46(11), 4587-4591 (1998)
Flavor compounds in straw mushrooms Volvariella volvacea harvested at different stages of maturity.
Mau JL, et al.
Journal of Agricultural and Food Chemistry, 45(12), 4726-4729 (1997)
Aroma characteristics of Neungee (Sarcodon aspratus).
Jeong OJ, et al.
Korean Journal of Food Science and Technology, 33(3), 307-312 (2001)
Cristian Gobbo et al.
ACS nano, 6(12), 10684-10698 (2012-12-12)
Controlling the molecular conformation of oligomers on surfaces through noncovalent interactions symbolizes an important approach in the bottom-up patterning of surfaces with nanoscale precision. Here we report on the design, synthesis, and scanning tunneling microscopy (STM) investigation of linear oligoamides
John A Arnott et al.
Expert opinion on drug discovery, 7(10), 863-875 (2012-09-21)
The role of lipophilicity in drug discovery and design is a critical one. Lipophilicity is a key physicochemical property that plays a crucial role in determining ADMET (absorption, distribution, metabolism, excretion, and toxicity) properties and the overall suitability of drug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service