Skip to Content
Merck

Skip To

1.08396

L-Tryptophan

EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, USP

Synonym(s):

L-Tryptophan, Trp

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
Molecular Weight:
204.23
UNSPSC Code:
12352209
EC Index Number:
200-795-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, ChP, JP, Ph. Eur., USP

product line

EMPROVE® EXPERT

form

solid

potency

16000 mg/kg LD50, oral (Rat)

pH

5.5-7.0 (20 °C, 10 g/L in H2O)

mp

290 °C (decomposition)

solubility

10 g/L

bulk density

400 kg/m3

application(s)

parenterals

storage temp.

2-30°C

SMILES string

[nH]1c2c(c(c1)CC(N)C(=O)O)cccc2

InChI

1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)

InChI key

QIVBCDIJIAJPQS-UHFFFAOYSA-N

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T2610000PHR11761.08374
application(s)

parenterals

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

agency

BP, JP, USP, ChP, Ph. Eur.

agency

-

agency

traceable to Ph. Eur. T2610000, traceable to USP 1700501

agency

-

form

solid

form

-

form

-

form

solid

product line

EMPROVE® EXPERT

product line

-

product line

-

product line

-

bulk density

400 kg/m3

bulk density

-

bulk density

-

bulk density

400 kg/m3

solubility

10 g/L

solubility

-

solubility

-

solubility

10 g/L


Still not finding the right product?

Explore all of our products under L-Tryptophan


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters