Skip to Content
Merck
All Photos(2)

Key Documents

H8520

Sigma-Aldrich

(±)-8-Hydroxy-2-(dipropylamino)tetralin hydrobromide

≥98%

Synonym(s):

(±)-2-Dipropylamino-8-hydroxy-1,2,3,4-tetrahydronaphthalene, (±)-8-Hydroxy-2-(dipropylamino)tetralin, (±)-8-OH-DPAT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H25NO · HBr
CAS Number:
Molecular Weight:
328.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

SMILES string

Br[H].CCCN(CCC)C1CCc2cccc(O)c2C1

InChI

1S/C16H25NO.BrH/c1-3-10-17(11-4-2)14-9-8-13-6-5-7-16(18)15(13)12-14;/h5-7,14,18H,3-4,8-12H2,1-2H3;1H

InChI key

BATPBOZTBNNDLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(±)-8-Hydroxy-2-(dipropylamino)tetralin hydrobromide is a potent and selective 5-hydroxytryptamine (5HT1A)-receptor agonist which is generally used as the standard in pharmacological studies.

Biochem/physiol Actions

Selective 5-HT1 agonist with high affinity for subtype 5-HT1A receptor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arylpiperazine agonists of the serotonin 5-HT 1A receptor preferentially activate cAMP signaling versus recruitment of β-arrestin-2.
Stroth N, et al.
Bioorganic & Medicinal Chemistry, 23(15), 4824-4830 (2015)
Laifu Li et al.
eLife, 10 (2021-06-04)
Consolation is a common response to the distress of others in humans and some social animals, but the neural mechanisms underlying this behavior are not well characterized. By using socially monogamous mandarin voles, we found that optogenetic or chemogenetic inhibition
8-Hydroxy-2-(di-n-propylamino)tetralin, a new centrally acting 5-hydroxytryptamine receptor agonist.
L E Arvidsson et al.
Journal of medicinal chemistry, 24(8), 921-923 (1981-08-01)
8-Hydroxy-2-(dipropylamino) tetralin, a new centrally acting 5-hydroxytryptamine receptor agonist.
Arvidsson L E, et al.
Journal of Medicinal Chemistry, 24(8), 921-923 (1981)
Improved preparation, chromatographic separation and X-ray crystallographic determination of the absolute configuration of the enantiomers of 8-hydroxy-2-(dipropylamino) tetralin (8-OH DPAT).
Karlsson A, et al.
Acta Chemica Scandinavica, 42, 231-236 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service