Skip to Content
Merck

Skip To

Q1004

Quipazine maleate salt

serotonin/5-hydroxytryptamine-2A/C receptor agonist

Quipazine maleate salt

Synonym(s):

2-(1-Piperazinyl)quinoline maleate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H15N3 · C4H4O4
CAS Number:
Molecular Weight:
329.35
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
227-314-2
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Quipazine maleate salt,

Quality Segment

solubility

H2O: soluble, alcohol: soluble, dilute aqueous acid: soluble

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O.C1CN(CCN1)c2ccc3ccccc3n2

InChI

1S/C13H15N3.C4H4O4/c1-2-4-12-11(3-1)5-6-13(15-12)16-9-7-14-8-10-16;5-3(6)1-2-4(7)8/h1-6,14H,7-10H2;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

QYJJDHZHSCTBII-BTJKTKAUSA-N

Application

Quipazine maleate salt has been used as a serotonin receptor agonist to study its effects on mouse models with spinal cord injury.[1][2] It has also been used as a serotonin receptor agonist to study its effects on the locomotion of zebrafish.[3]

Biochem/physiol Actions

Quipazine belongs to the class of psychedelics[4] and acts as a serotonin/5-hydroxytryptamine-2A/C receptor agonist.[2] It also acts as an inhibitor of prolactin.[5]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Q1250F8927P9233
Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

2-8°C

solubility

H2O: soluble, dilute aqueous acid: soluble, alcohol: soluble

solubility

H2O: slightly soluble 1.2 mg/mL, ethanol: 8 mg/mL

solubility

DMSO: soluble >20 mg/mL, H2O: insoluble

solubility

DMSO: >10 mg/mL, H2O: >10 mg/mL

Gene Information

human ... HTR1A(3350), HTR1B(3351), HTR1D(3352), HTR1E(3354), HTR1F(3355), HTR2A(3356), HTR2B(3357), HTR2C(3358), HTR3A(3359), HTR3B(9177), HTR3C(170572), HTR3D(200909), HTR3E(285242), HTR4(3360), HTR5A(3361), HTR5B(645694), HTR6(3362), HTR7(3363)

Gene Information

-

Gene Information

-

Gene Information

human ... DRD3(1814)


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

Active Filters

  1. 1 Ratings-only review