Skip to Content
Merck
All Photos(1)

Key Documents

H16752

Sigma-Aldrich

Humic acid sodium salt

technical grade

Synonym(s):

Humic acid Na salts, Sodium humates, Sodium salts humic acids

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

grade

technical grade

mp

>300 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

Humic acid possess excellent complexing ability with heavy metal. Humic acid sodium salt acts as a proxy for humic-like substances that were identified as a major component of the isolated organic matter in the atmospheric aerosol.

Humic acid sodium salt, also known as sodium humate is used to synthesize superabsorbents.

Application

Humic acid sodium salt is suitable reagent used to study the hygroscopic growth of solid aerosol particles consisting of mixtures of ammonium sulfate, adipic acid and humic acid mixtures. It is suitable reagent used to study the phase transitions and hygroscopic growth of humic acid aerosols by Fourier transform infra-red (FTIR) spectroscopy and tandem differential mobility analysis (TDMA).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hygroscopic growth and water uptake kinetics of two-phase aerosol particles consisting of ammonium sulfate, adipic and humic acid mixtures.
Sjogren S, et al.
Journal of Aerosol Science, 38(2), 151-171 (2007)
V Kuokkanen et al.
Water research, 79, 79-87 (2015-05-15)
Electrocoagulation (EC) treatment of 100 mg/L synthetic wastewater (SWW) containing humic acids was optimized (achieving 90% CODMn and 80% DOC removal efficiencies), after which real peat bog drainage waters (PBDWs) from three northern Finnish peat bogs were also treated. High pollutant
Tangui Léziart et al.
Environmental technology, 40(13), 1734-1743 (2019-02-20)
Chlorine is globally the most widely used chemical for water disinfection. Whereas disinfection efficiency is well known to depend on water pH and temperature, the effect of turbidity is less well studied. Although turbidity is measured online in most drinking
Qi Zhao et al.
Chemosphere, 256, 127021-127021 (2020-05-22)
The effect of gamma-irradiation doses of 0, 10, 100, and 500 kGy at the dose rates of 1 or 0.1 kGy/h on the molecular and chemical properties of humic substances (HS) were investigated using total organic carbon measurements, UV-Vis spectrometry, 13C nuclear magnetic
Synthesis and characterization of chitosan-g-poly (acrylic acid)/sodium humate superabsorbent
Liu J, et al.
Carbohydrate Polymers, 70, 166-173 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service