Skip to Content
Merck
All Photos(1)

Documents

53680

Sigma-Aldrich

Humic acid

technical

Synonym(s):

PGS 10

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

grade

technical

Quality Level

form

solid

ign. residue

~20%

InChI

1S/K

InChI key

ZLMJMSJWJFRBEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Humic acid (HA) is a heterogeneous macromolecule mainly found in soil and water.

Application

Humic acid has been used to prepare stock solution employed in the following experiments:
  • Coagulation activity assay of Moringa oleifera protein extracts for humic acid removal.
  • Adsorption edge experiment to study its effect on the sorption of tetracycline onto goethite.
  • Evaluation of the ability of polymeric adsorbent Supelite Dax-8 to remove humic acid from nucleic acid extracts prior to quantitative real-time PCR (qPCR).

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improving qPCR efficiency in environmental samples by selective removal of humic acids with DAX-8.
Schriewer A, et al.
Journal of Microbiological Methods, 85(1), 16-21 (2011)
Nuria Polo-Cavia et al.
Aquatic toxicology (Amsterdam, Netherlands), 172, 30-35 (2016-01-15)
Recent studies suggest that direct mortality and physiological effects caused by pollutants are major contributing factors to global amphibian decline. However, even sublethal concentrations of pollutants could be harmful if they combined with other factors to cause high mortality in
Joon-Yung Cha et al.
Scientific reports, 10(1), 15042-15042 (2020-09-16)
Humic acid (HA) is composed of a complex supramolecular association and is produced by humification of organic matters in soil environments. HA not only improves soil fertility, but also stimulates plant growth. Although numerous bioactivities of HA have been reported
Bo Højris et al.
Scientific reports, 6, 23935-23935 (2016-04-05)
Today, microbial drinking water quality is monitored through either time-consuming laboratory methods or indirect on-line measurements. Results are thus either delayed or insufficient to support proactive action. A novel, optical, on-line bacteria sensor with a 10-minute time resolution has been
Adsorption of tetracycline onto goethite in the presence of metal cations and humic substances.
Zhao Y, et al.
Journal of Colloid and Interface Science, 361(1), 247-251 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service