Recommended Products
sterility
non-sterile
form
powder
manufacturer/tradename
NutriSelect® Basic
technique(s)
microbiological culture: suitable
application(s)
agriculture
environmental
food and beverages
life science and biopharma
suitability
marine bacteria
General description
Sea salts are unrefined salt, which is harvested from ocean and saltwater deposit. It is used to maintain the body′s salt balance.
Application
Sea salts has been used:
- for the preparation of artificial seawater to culture microalgae
- to assess nitrogen fixation rates
- as a supplement in Hugh and Leifson′s oxidation fermentation (OF) basal medium
Usually used at approx. 40 g/L for culturing marine bacteria.
Other Notes
An artificial salt mixture closely resembling the composition of the dissolved salts of ocean water.
Preparation Note
Usually used at ~ 40 g/L for culturing marine bacteria.
After dissolving, adjust pH to 8.2 using 0.1 N solution of sodium hydroxide or hydrochloric acid.
After dissolving, adjust pH to 8.2 using 0.1 N solution of sodium hydroxide or hydrochloric acid.
Footnote
We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Shunichi Takahashi et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(11), 4203-4208 (2008-03-07)
Coral bleaching, caused by heat stress, is accompanied by the light-induced loss of photosynthetic pigments in in situ symbiotic dinoflagellate algae (Symbiodinium spp.). However, the molecular mechanisms responsible for pigment loss are poorly understood. Here, we show that moderate heat
Sea Salt Sweet: The Art of Using Salts for the Ultimate Dessert Experience (2015)
The Body Ecology Diet (2011)
Maria Mansson et al.
Marine drugs, 9(12), 2537-2552 (2012-03-01)
During a global research expedition, more than five hundred marine bacterial strains capable of inhibiting the growth of pathogenic bacteria were collected. The purpose of the present study was to determine if these marine bacteria are also a source of
P Zieger et al.
Nature communications, 8, 15883-15883 (2017-07-04)
Sea spray is one of the largest natural aerosol sources and plays an important role in the Earth's radiative budget. These particles are inherently hygroscopic, that is, they take-up moisture from the air, which affects the extent to which they
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service