D215554
Dipropylene glycol
99%, mixture of isomers
Synonym(s):
DPG, Bis(hydroxypropyl) ether
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOC3H6OC3H6OH
CAS Number:
Molecular Weight:
134.17
Beilstein:
1698372
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
4.6 (vs air)
Quality Level
vapor pressure
<0.01 mmHg ( 20 °C)
Assay
99%
form
liquid
refractive index
n20/D 1.441 (lit.)
density
1.023 g/mL at 25 °C (lit.)
SMILES string
CC(O)COCC(C)O.CC(O)COC(C)CO.CC(CO)OC(C)CO
InChI
1S/C6H14O3/c1-3-5(7)9-6(8)4-2/h5-8H,3-4H2,1-2H3
InChI key
QYSXYAURTRCDJU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Dipropylene glycol can be used as a solvent in the TEMPO-mediated living radical dispersion polymerization of styrene.
Packaging
Packaged in plastic bottles
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
266.0 °F - closed cup
Flash Point(C)
130 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Solvent effect on TEMPO-mediated living free radical dispersion polymerization of styrene.
Shim S E, et al.
Polymer, 45(14), 4731-4739 (2004)
Patricia Carlisle et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 107(5), 1491-1503 (2018-09-29)
At least 26% of recent battlefield injuries are to the craniomaxillofacial (CMF) region. Recombinant human bone morphogenetic protein 2 (rhBMP-2) is used to treat CMF open fractures, but several complications have been associated with its use. This study tested the
Paulina Kosmela et al.
Materials (Basel, Switzerland), 13(5) (2020-03-14)
The paper describes the preparation and characterization of rigid polyurethane-polyisocyanurate (PUR-PIR) foams obtained with biopolyol synthesized in the process of liquefaction of biomass from the Baltic Sea. The obtained foams differed in the content of biopolyol in polyol mixture (0-30
Mónica Bueno et al.
Food chemistry, 281, 49-56 (2019-01-20)
This work presents a headspace-solid phase microextraction-gas chromatography-mass spectrometry (HS-SPME-GC-MS) method for the analysis of solid food samples in extended experiments. The final procedure was used to quantify 30 volatile compounds in fresh beef. The strategy adds robustness to the
Remodeling of injectable, low-viscosity polymer/ceramic bone grafts in a sheep femoral defect model.
Anne D Talley et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 105(8), 2333-2343 (2016-08-11)
Ceramic/polymer composite bone grafts offer the potential advantage of combining the osteoconductivity of ceramic component with the ductility of polymeric component, resulting in a graft that meets many of the desired properties for bone void fillers (BVF). However, the relative
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service