Skip to Content
Merck
All Photos(1)

Key Documents

597236

Sigma-Aldrich

Di(propylene glycol) dibenzoate

75%, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CO2C3H6)2O
CAS Number:
Molecular Weight:
342.39
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

75%

refractive index

n20/D 1.528 (lit.)

bp

232 °C/5 mmHg (lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

O=C(OCCCOCCCOC(=O)c1ccccc1)c2ccccc2

InChI

1S/C20H22O5/c21-19(17-9-3-1-4-10-17)24-15-7-13-23-14-8-16-25-20(22)18-11-5-2-6-12-18/h1-6,9-12H,7-8,13-16H2

InChI key

BYQDGAVOOHIJQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Di(propylene glycol) dibenzoate (DPGDB) is a widely used plasticizer that has ether linkages linked with two benzoate groups.

Application

DPGDB may be used as a diluent for the preparation of polysulfone membranes by heat induced phase separation. It finds potential applications in water treatment and food processing. It may also be used as a plasticizer with poly(vinyl) chloride (PVC) for the fabrication of diamond coated PVC.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

377.6 °F - closed cup

Flash Point(C)

192 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication of polysulfone membrane via thermally induced phase separation process.
Liu Z, et al.
Materials Letters, 195(6), 190-193 (2017)
Development of new PVC materials. Characterization and feasibility of diamond coatings on model PVC materials.
Koponen HK, et al.
Diamond and Related Materials, 16(3), 630-636 (2007)
Designing green plasticizers: Linear alkyl diol dibenzoate plasticizers and a thermally reversible plasticizer
Erythropel H, et al.
Polymers, 10(6), 646-646 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service