Skip to Content
Merck
All Photos(1)

Documents

484253

Sigma-Aldrich

Di(propylene glycol) methyl ether, mixture of isomers

≥99%

Synonym(s):

DOWANOL® DPM, Dipropylene glycol monomethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC3H6OC3H6OH
CAS Number:
Molecular Weight:
148.20
EC Number:
UNSPSC Code:
12162002
NACRES:
NA.23

vapor pressure

0.4 mmHg ( 25 °C)

Assay

≥99%

refractive index

n20/D 1.422 (lit.)

bp

190 °C (lit.)

density

0.951 g/mL at 25 °C

InChI

1S/C7H16O3/c1-6(4-8)10-5-7(2)9-3/h6-8H,4-5H2,1-3H3

InChI key

CUDYYMUUJHLCGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Di(propylene glycol) methyl ether is an organic solvent used in various applications such as ceramic ink preparation, etching, and bioactive glass suspension.

Legal Information

Product of Dow Chemical
DOWANOL is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formulation of a Ceramic Ink for 3D Inkjet Printing
Dennis Graf, et al.
Micromachines, 12 (2021)
Bioactive glass suspensions preparation for suspension plasma spraying
E.Ca?as, et al
J. Eur. Ceram. Soc., 36, 4281-4290 (2016)
Alternative materials for printed circuit board production: An environmental perspective
Mohammad Naji Nassajfar, et al.
Sustainability, 13, 12126-12126 (2021)
Michael Koontz et al.
International journal of toxicology, 25(2), 95-107 (2006-04-07)
Computer modeling of aggregate exposure provides the capability to estimate the range of doses that can occur from product use and to understand the relative importance of different routes of exposure. This paper presents an assessment of aggregate occupational exposure
Toshiaki Tsukatani et al.
Chemical senses, 28(1), 25-32 (2002-12-28)
Detection thresholds are typically obtained by presenting a subject with serial dilutions of an odorant. Many factors, including the solvent used to dilute the odorant, can influence the measurement of detection thresholds. Differences have been reported in detection thresholds for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service