Skip to Content
Merck
All Photos(1)

Documents

B9009

Sigma-Aldrich

Benzo[ghi]perylene

98%

Synonym(s):

1,12-Benzoperylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33
Beilstein:
1913029
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

solid

bp

>500 °C (lit.)

mp

277-279 °C (lit.)

SMILES string

c1cc2ccc3ccc4ccc5cccc6c(c1)c2c3c4c56

InChI

1S/C22H12/c1-3-13-7-9-15-11-12-16-10-8-14-4-2-6-18-17(5-1)19(13)21(15)22(16)20(14)18/h1-12H

InChI key

GYFAGKUZYNFMBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

An n-channel organic semiconductor.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Py and BPe solvent polarity scales: effect of temperature on pyrene and benzo[ghi]perylene fluorescence spectra.
R Waris et al.
The Analyst, 113(9), 1465-1467 (1988-09-01)
S H Cherng et al.
Toxicology and applied pharmacology, 170(1), 63-68 (2001-01-06)
Although benzo[g,h,i]perylene (BghiP) has been found to promote the carcinogenesis of benzo[a]pyrene (BaP) in animal models, not much is known about this cocarcinogenic mechanism. In this study, human hepatoma HepG2 cells cotreated with BaP and BghiP were used as a
Nobuyasu Itoh et al.
Analytica chimica acta, 615(1), 47-53 (2008-04-29)
Microwave-assisted extraction using 1M KOH/methanol (alkaline-MAE) in combination with solid-phase extraction treatment was developed and applied to polycyclic aromatic hydrocarbons (PAHs) in a sediment sample. Although various conditions were examined (100 or 150 degrees C for 10 or 30 min)
Karl L Platt et al.
Chemical research in toxicology, 18(4), 700-710 (2005-04-19)
Carcinogenic polycyclic aromatic hydrocarbons (PAH), e.g., benzo[a]pyrene (BaP), possess a bay region comprising an ortho-fused benzene ring. Benzo[ghi]perylene (BghiP) represents the group of PAHs lacking such a "classic" bay region and hence cannot be metabolically converted like BaP to bay
Stepan Boitsov et al.
Marine environmental research, 68(5), 236-245 (2009-08-01)
Polycyclic aromatic hydrocarbons (PAH) were measured in sediment cores from 13 locations in South-Western Barents Sea as part of a detailed study of the Norwegian seabed under the MAREANO program. The generally low PAH levels found, an average around 200

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service