Skip to Content
Merck
All Photos(1)

Documents

275336

Sigma-Aldrich

Benzo[b]fluoranthene

98%

Synonym(s):

2,3-Benzfluoranthene, 3,4-Benz[e]acephenanthrylene, 3,4-Benzfluoranthene, 3,4-Benzofluoranthene, Benz[e]acephenanthrylene, Benzo[e]fluoranthene, NSC 89265

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

163-165 °C (lit.)

SMILES string

c1ccc-2c(c1)-c3cccc4c3c-2cc5ccccc45

InChI

1S/C20H12/c1-2-7-14-13(6-1)12-19-16-9-4-3-8-15(16)18-11-5-10-17(14)20(18)19/h1-12H

InChI key

FTOVXSOBNPWTSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The oxidative damage and bioaccumulation of juvenile scallops Chlamys farreri exposed to benzo[b]fluoranthene, a polycyclic aromatic hydrocarbon, was studied.

Benz[e]acephenanthrylene is a polycyclic aromatic hydrocarbon composed of a central 5-membered ring surrounded by four benzene rings.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shangbo Zhou et al.
The Science of the total environment, 763, 143030-143030 (2020-11-02)
Bioanalytical equivalents (BEQs) of mixtures and environmental samples are widely used to reflect the potential threat of pollutants in the environment and can be obtained by bioassays or using chemical analysis combined with relative potencies (REPs). In this study, the
Meng Xiu et al.
Ecotoxicology and environmental safety, 107, 103-110 (2014-06-27)
The study is aimed at investigating the bioaccumulation and oxidative damage of juvenile scallops (Chlamys farreri) exposed to three selected PAHs: benzo[a]pyrene (BaP), benzo[b]fluoranthene (BbF) and chrysene (CHR). For this purpose, a study was performed on juvenile scallops exposed to
Polycyclic aromatic hydrocarbons determination
Gachanja AN and Maritim PK
Encyclopedia of Analytical Science, 328-340 (2019)
S Nesnow et al.
Environmental health perspectives, 101 Suppl 3, 37-42 (1993-10-01)
DNA-carcinogen adducts offer a potential dosimeter for environmental genotoxicants reaching the exposed individual. Because the target tissues for many chemical carcinogens are not readily accessible for monitoring adducts in humans, peripheral blood lymphocytes (PBLs) have served as surrogate sources of
S Aubin et al.
Journal of the Air & Waste Management Association (1995), 50(12), 2093-2101 (2001-01-05)
Benzo[b]fluoranthene (B[b]F) was used in relative abundance ratios (RARs), a parameter obtained by dividing the concentration of individual polycyclic aromatic hydrocarbons (PAHs) found in a given sample by the concentration of B[b]F in the same sample. The B[b]F RARs were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service