Skip to Content
Merck
All Photos(1)

Key Documents

416703

Sigma-Aldrich

Acenaphthylene

99%

Synonym(s):

Cyclopenta[de]naphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8
CAS Number:
Molecular Weight:
152.19
Beilstein:
774092
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

280 °C (lit.)

mp

78-82 °C (lit.)

density

0.899 g/mL at 25 °C (lit.)

SMILES string

c1cc2C=Cc3cccc(c1)c23

InChI

1S/C12H8/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-8H

InChI key

HXGDTGSAIMULJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acenaphthylene has been used to investigate the photodimerization of acenaphthylene in micellar and hydrogel media.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

251.6 °F - closed cup

Flash Point(C)

122.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An electron spin resonance study of the carbonization of the aromatic hydrocarbon acenaphthylene.
Singer LS and Lewis IC.
Carbon, 2(2), 115-120 (1964)
Durairajan Madhavan et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 2(2), 95-97 (2003-04-01)
The photochemical dimerisation of acenaphthylene in the presence of various cation-exchanged bentonite clays has been studied. While the cis-dimer is the predominant product when smaller cations are present in the clay interlayer, the presence of heavier atoms in the clay
Kinetics and products of the gas-phase reactions of acenaphthene with hydroxyl radicals, nitrate radicals and ozone.
Zhou S and Wenger JC.
Atmospheric Environment, 72, 97-104 (2013)
M J Schocken et al.
Applied and environmental microbiology, 48(1), 10-16 (1984-07-01)
A Beijerinckia sp. and a mutant strain, Beijerinckia sp. strain B8/36, were shown to cooxidize the polycyclic aromatic hydrocarbons acenaphthene and acenaphthylene. Both organisms oxidized acenaphthene to the same spectrum of metabolites, which included 1-acenaphthenol, 1-acenaphthenone, 1,2-acenaphthenediol, acenaphthenequinone, and a
Philipp Eyring et al.
Journal of separation science, 44(2), 600-608 (2020-11-14)
Food can contain unwanted compounds and need to be analyzed for compounds like carcinogenic polycyclic aromatic hydrocarbons to ensure consumer safety. The analytes need to be extracted from the sample matrix and cleaned-up to enable detection. However, established methods for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service