Skip to Content
Merck
All Photos(2)

Key Documents

B2209

Sigma-Aldrich

Benz[a]anthracene

99%

Synonym(s):

1,2-Benzanthracene, Tetraphene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
Beilstein:
1909298
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

bp

437.6 °C (lit.)

mp

157-159 °C (lit.)

SMILES string

c1ccc2cc3c(ccc4ccccc34)cc2c1

InChI

1S/C18H12/c1-2-7-15-12-18-16(11-14(15)6-1)10-9-13-5-3-4-8-17(13)18/h1-12H

InChI key

DXBHBZVCASKNBY-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Application

Benz[a]anthracene can be used in the synthesis of other polycyclic aromatic hydrocarbons such as tribenzo[a,c,f]tetraphene.2 It can also be used for phosphorescence applications.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal induced phosphorescence from Benz (a) anthracene microcrystals at room temperature.
Maity S, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 157, 61-68 (2016)
Direct arylation of polycyclic aromatic hydrocarbons through palladium catalysis.
Mochida K, et al.
Journal of the American Chemical Society, 133(28), 10716-10719 (2011)
Synthesis, Structural and Spectroscopic Characterization of Four [(η6?PAH) Cr (CO)3] Complexes (PAH= Pyrene, Perylene, Chrysene, 1, 2?Benzanthracene).
Arrais A, et al.
European Journal of Inorganic Chemistry, 2004(7), 1505-1513 (2004)
Philipp Eyring et al.
Journal of separation science, 44(2), 600-608 (2020-11-14)
Food can contain unwanted compounds and need to be analyzed for compounds like carcinogenic polycyclic aromatic hydrocarbons to ensure consumer safety. The analytes need to be extracted from the sample matrix and cleaned-up to enable detection. However, established methods for
Xin-Chun Zhao et al.
EBioMedicine, 53, 102689-102689 (2020-03-03)
How the oncoprotein epidermal growth factor receptor (EGFR) evades proteolytic degradation and accumulates in non-small cell lung cancer (NSCLC) remains unclear, and ubiquitin pathway genes (UPGs) that are critical to NSCLC needs to be systematically identified. A total of 696

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service