Skip to Content
Merck
All Photos(1)

Key Documents

906328

Sigma-Aldrich

BTTAA

≥95%

Synonym(s):

2-(4-((Bis((1-(tert-butyl)-1H-1,2,3-triazol-4-yl)methyl)amino)methyl)-1H-1,2,3-triazol-1-yl)acetic acid, Copper click-chemistry ligand, Water-soluble CuAAC ligand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30N10O2
CAS Number:
Molecular Weight:
430.51
MDL number:
UNSPSC Code:
12352200

Assay

≥95%

form

solid

reaction suitability

reaction type: click chemistry

availability

available only in USA

storage temp.

2-8°C

SMILES string

[n]1(nnc(c1)CN(Cc3nn[n](c3)C(C)(C)C)Cc2nn[n](c2)CC(=O)O)C(C)(C)C

InChI

1S/C19H30N10O2/c1-18(2,3)28-11-15(21-24-28)8-26(7-14-10-27(23-20-14)13-17(30)31)9-16-12-29(25-22-16)19(4,5)6/h10-12H,7-9,13H2,1-6H3,(H,30,31)

InChI key

MGQYHUDOWOGSQI-UHFFFAOYSA-N

Application

BTTAA is a next-generation, water-soluble ligand for the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) that dramatically accelerates reaction rates and suppresses cell cytotoxicity. The biocompatibility and fast kinetics of BTTAA are advancements from water-insoluble TBTA and are desirable for bio conjugation in diverse chemical biology experiments.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maiyun Yang et al.
Angewandte Chemie (International ed. in English), 51(31), 7674-7679 (2012-07-06)
Live-cell pH measurements: An environment-sensitive fluorophore (green) was site-specifically introduced on HdeA, an acid-resistant chaperone showing pH-mediated conformational changes under low pH conditions. A survey of the attachment sites led to the discovery of one position on HdeA at which
Yongxian Xu et al.
Angewandte Chemie (International ed. in English), 57(15), 3949-3953 (2018-02-14)
Membrane voltage is an important biophysical signal that underlies intercellular electrical communications. A fluorescent voltage indicator is presented that enables the investigation of electrical signaling at high spatial resolution. The method is built upon the site-specific modification of microbial rhodopsin
Fast, cell-compatible click chemistry with copper-chelating azides for biomolecular labeling.
Chayasith Uttamapinant et al.
Angewandte Chemie (International ed. in English), 51(24), 5852-5856 (2012-05-05)
Samra Obeid et al.
Chemical communications (Cambridge, England), 48(67), 8320-8322 (2012-07-07)
Modified nucleotides play a paramount role in many cutting-edge biomolecular techniques. The present structural study highlights the plasticity and flexibility of the active site of a DNA polymerase while incorporating non-polar "Click-able" nucleotide analogs and emphasizes new insights into rational
Yinliang Yang et al.
Molecules (Basel, Switzerland), 18(10), 12599-12608 (2013-10-16)
Activity-based protein profiling uses chemical probes that covalently attach to active enzyme targets. Probes with conventional tags have disadvantages, such as limited cell permeability or steric hindrance around the reactive group. A tandem labeling strategy with click chemistry is now

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service