906301
BTTES
Synonym(s):
3-(4-((Bis((1-(tert-butyl)-1H-1,2,3-triazol-4-yl)methyl)amino)methyl)-1H-1,2,3-triazol-1-yl)propane-1-sulfonic acid, Copper click-chemistry ligand, Water-soluble CuAAC ligand
About This Item
Recommended Products
form
solid
reaction suitability
reaction type: click chemistry
availability
available only in USA
storage temp.
2-8°C
SMILES string
[S](=O)(=O)(O)CCC[n]1nnc(c1)CN(Cc3nn[n](c3)C(C)(C)C)Cc2nn[n](c2)C(C)(C)C
InChI
1S/C20H34N10O3S/c1-19(2,3)29-14-17(22-25-29)11-27(12-18-15-30(26-23-18)20(4,5)6)10-16-13-28(24-21-16)8-7-9-34(31,32)33/h13-15H,7-12H2,1-6H3,(H,31,32,33)
InChI key
WMEZDVILBKIODK-UHFFFAOYSA-N
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Self-react. C
Storage Class Code
5.2 - Organic peroxides and self-reacting hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service