Skip to Content
Merck
All Photos(3)

Documents

762342

Sigma-Aldrich

Tris(3-hydroxypropyltriazolylmethyl)amine

95%

Synonym(s):

3,3′,3′′-(4,4′,4′′-(Nitrilotris(methylene))tris(1H-1,2,3-triazole-4,1-diyl))tris(propan-1-ol), THPTA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H30N10O3
CAS Number:
Molecular Weight:
434.50
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

reaction suitability

reaction type: click chemistry

mp

331-336 °C (decomposition)

storage temp.

2-8°C

SMILES string

OCCCN1N=NC(CN(CC2=CN(CCCO)N=N2)CC3=CN(CCCO)N=N3)=C1

InChI

1S/C18H30N10O3/c29-7-1-4-26-13-16(19-22-26)10-25(11-17-14-27(23-20-17)5-2-8-30)12-18-15-28(24-21-18)6-3-9-31/h13-15,29-31H,1-12H2

InChI key

VAKXPQHQQNOUEZ-UHFFFAOYSA-N

Application

Effective ligand for copper-catalyzed azide-alkyne cycloadditions (CuAAC).
Tris(3-hydroxypropyltriazolylmethyl)amine is a water-soluble copper(I) stabilizing ligand mainly used in copper(I)-catalyzed alkyne-azide cycloaddition (CuAAC) reactions.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vu Hong et al.
Bioconjugate chemistry, 21(10), 1912-1916 (2010-10-05)
The copper-catalyzed azide-alkyne cycloaddition (CuAAC) reaction, optimized for biological molecules in aqueous buffers, has been shown to rapidly label mammalian cells in culture with no loss in cell viability. Metabolic uptake and display of the azide derivative of N-acetylmannosamine developed
Sheng Wang et al.
The Journal of pathology, 243(1), 78-88 (2017-06-29)
Protein modification by O-linked β-N-acetylglucosamine (O-GlcNAc) is emerging as an important factor in the pathogenesis of sporadic Alzheimer's disease (AD); however, detailed molecular characterization of this important protein post-translational modification at the proteome level has been highly challenging, owing to
Patrik Fladischer et al.
Biotechnology journal, 14(3), e1800125-e1800125 (2018-06-05)
The site-specific incorporation of non-canonical amino acids (ncAAs) at amber codons requires an aminoacyl-tRNA synthetase and a cognate amber suppressor tRNA (tRNACUA ). The archaeal tyrosyl-tRNA synthetase from Methanocaldococcus jannaschii and the pyrrolysyl-tRNA synthetase (PylRS) from Methanosarcina mazei have been
A two-component'double-click'approach to peptide stapling.
Lau YH, et al.
Nature Protocols, 10(4), 585-585 (2015)
Tingbi Zhao et al.
Nanoscale, 9(28), 9841-9847 (2017-05-10)
A large amount of proteins are post-translationally modified with a sialic acid terminal oligosaccharide, and sialylation directly affects the function of glycoproteins and adjusts relevant biological processes. Herein, we developed a method for imaging analysis of protein-specific sialylation on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service