Skip to Content
Merck
All Photos(1)

Key Documents

PHR1279

Supelco

Sodium ascorbate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(+)-Sodium L-ascorbate, L(+)-Ascorbic acid sodium salt, Vitamin C sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NaO6
CAS Number:
Molecular Weight:
198.11
Beilstein:
3767246
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000039
traceable to USP 1613509

API family

sodium ascorbate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

220 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[Na+].OC[C@H](O)[C@H]1OC(=O)C(O)=C1[O-]

InChI

1S/C6H8O6.Na/c7-1-2(8)5-3(9)4(10)6(11)12-5;/h2,5,7-10H,1H2;/q;+1/p-1/t2-,5+;/m0./s1

InChI key

PPASLZSBLFJQEF-RXSVEWSESA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium ascorbate, also known as E301, is a sodium salt of ascorbic acid. It is generally used as an antioxidant in pharmaceutical formulations and food products.
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards and pharmacopeia primary standards.

Application

Sodium ascorbate may be used as a pharmaceutical reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by titrimetry and liquid chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC1682 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium Ascorbate
Handbook of Pharmaceutical Excipients 6th edition Pharmaceutical Press, 625-627 (2009)
Sodium Ascorbate
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 6997-6998 (2019)
Sodium Ascorbate
Pharmacopeia, US
United States Pharmacopeia/National Formulary, 35(6), 4018-4018 (2018)
Aleksander Hinek et al.
Journal of dermatological science, 75(3), 173-182 (2014-07-13)
Vitamin C (L-ascorbic acid), a known enhancer of collagen deposition, has also been identified as an inhibitor of elastogenesis. Present studies explored whether and how the L-ascorbic acid derivative (+) sodium L-ascorbate (SA) would affect production of collagen and elastic
H R Harris et al.
British journal of cancer, 109(1), 257-264 (2013-06-06)
Vitamin C may influence cancer progression through its antioxidant properties. However, the evidence from observational epidemiologic studies on vitamin C intake and survival following breast cancer diagnosis is not consistent, and the safety of vitamin C supplements following breast cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service