Merck
  • The effect of amorphous pyrophosphate on calcium phosphate cement resorption and bone generation.
The effect of amorphous pyrophosphate on calcium phosphate cement resorption and bone generation.
Biomaterials (2013-06-12)
Liam M Grover, Adrian J Wright, Uwe Gbureck, Aminat Bolarinwa, Jiangfeng Song, Yong Liu, David F Farrar, Graeme Howling, John Rose, Jake E Barralet
ABSTRACT
Pyrophosphate ions are both inhibitors of HA formation and substrates for phosphatase enzymes. Unlike polyphosphates their hydrolysis results simultaneously in the complete loss of mineral formation inhibition and a localised elevation in orthophosphate ion concentration. Despite recent advances in our knowledge of the role of the pyrophosphate ion, very little is known about the effects of pyrophosphate on bone formation and even less is known about its local delivery. In this work we first developed a self setting pyrophosphate based calcium cement system with appropriate handling properties and then compared its in vivo degradation properties with those of a non-pyrophosphate containing control. Contrary to expectation, the presence of the pyrophosphate phase in the cement matrix did not inhibit mineralisation of the healing bone around the implant, but actually appeared to stimulate it. In vitro evidence suggested that enzymatic action accelerated dissolution of the inorganic pyrophosphate ions, causing a simultaneous loss of their mineralisation inhibition and a localised rise in supersaturation with respect to HA. This is thought to be a rare example of a biologically responsive inorganic material and these materials seem to be worthy of further investigation. Bioceramics to date have mainly been limited to orthophosphate, silicate and carbonate salts of calcium, here we report the successful application of a pyrophosphate material as a degradable osteoconductive bone repair cement.
Sigma-Aldrich
Hydroxyapatite, reagent grade, powder, synthetic
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Calcium phosphate tribasic, suitable for plant cell culture, BioReagent, powder
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis