919101
Hydroxyapatite, acicular powder
≥96%, 60 nm±10 nm particle size
Synonym(s):
Calcium hydroxyphosphate, Calcium phosphate tribasic, HA, HaP, Hydroxylapatite, Tribasic calcium phosphate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Ca10(PO4)6(OH)2
UNSPSC Code:
12352305
NACRES:
NA.23
Recommended Products
Application
Hydroxyapatite is a naturally occurring mineral of biological importance, due to its similarity in size, crystallography and composition to human hard tissue. Human bone morphology is mostly acicular hydroxyapatite, which makes it an ideal material for bone and dental applications.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
In Situ Precipitation of Cluster and Acicular Hydroxyapatite onto Porous Poly(γ-benzyl-l-glutamate) Microcarriers for Bone Tissue Engineering
Bu S, et al.
ACS Applied Materials & Interfaces, 12(11), 12468-12477 (2020)
Three common preparation methods of hydroxyapatite.
Ma G, et al.
IOP Conference Series: Materials Science and Engineering, 688, 033057-033057 (2019)
Margarida M Fernandes et al.
ACS applied materials & interfaces, 11(48), 45265-45275 (2019-11-05)
Bone tissue repair strategies are gaining increasing relevance due to the growing incidence of bone disorders worldwide. Biochemical stimulation is the most commonly used strategy for cell regeneration, while the application of physical cues, including magnetic, mechanical, or electrical fields
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service