Skip to Content
Merck
All Photos(3)

Key Documents

642045

Sigma-Aldrich

Cadmium nitrate tetrahydrate

98%

Synonym(s):

Nitric acid, cadmium salt tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cd(NO3)2 · 4H2O
CAS Number:
Molecular Weight:
308.48
Beilstein:
8156730
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
98%
form:
solid

Quality Level

Assay

98%

form

solid

reaction suitability

reagent type: catalyst
core: cadmium

mp

59.5 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+](=O)O[Cd]O[N+]([O-])=O

InChI

1S/Cd.2NO3.4H2O/c;2*2-1(3)4;;;;/h;;;4*1H2/q+2;2*-1;;;;

InChI key

QOYRNHQSZSCVOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Target Organs

Kidney,Bone

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M S Monteiro et al.
Ecotoxicology and environmental safety, 72(3), 811-818 (2008-10-28)
Laboratory and field studies have provided encouraging insights into the capacity of plants to act as biomonitors of environmental quality through the use of biomarkers. However, a better understanding of the overall process of Cd-induced senescence, describing the cascade of
Akmal Muhammad et al.
Chemosphere, 60(4), 508-514 (2005-06-14)
A study was conducted to evaluate the effects of different concentrations of lead (Pb) and cadmium (Cd) applied as their nitrates on soil microbial biomass carbon (C(mic)) and nitrogen (N(mic)), and substrate utilization pattern of soil microbial communities. The C(mic)
G De Boeck et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 155(4), 503-508 (2010-01-02)
Recent studies indicate that elasmobranch fish respond differently to metal exposure than marine teleosts. Accumulation rates can be high, which despite the fact that normal background levels for metals in the marine environment are low, is worrying due to the
Ralph Blum et al.
The Plant journal : for cell and molecular biology, 49(4), 740-749 (2007-01-27)
Detoxification of xenobiotic compounds and heavy metals is a pivotal capacity of organisms, in which glutathione (GSH) plays an important role. In plants, electrophilic herbicides are conjugated to the thiol group of GSH, and heavy metal ions form complexes as
E Helene Larsson et al.
Journal of experimental botany, 53(368), 447-453 (2002-02-16)
In order to test the potential effect of prior exposure to different Cd concentrations on Cd uptake and accumulation, plants of Arabidopsis thaliana, including a phytochelatin-deficient mutant, cad1-3, and the wild type, were compared. For Cd uptake experiments, plants were

Articles

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service