Skip to Content
Merck
All Photos(3)

Key Documents

201073

Sigma-Aldrich

Aluminum-tri-sec-butoxide

97%

Synonym(s):

Aluminum sec-butoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al[OCH(CH3)C2H5]3
CAS Number:
Molecular Weight:
246.32
Beilstein:
3910908
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

autoignition temp.

763 °F

expl. lim.

9.8 %

reaction suitability

core: aluminum

bp

200-206 °C/30 mmHg (lit.)

density

0.967 g/mL at 25 °C (lit.)

SMILES string

CCC(C)O[Al](OC(C)CC)OC(C)CC

InChI

1S/3C4H9O.Al/c3*1-3-4(2)5;/h3*4H,3H2,1-2H3;/q3*-1;+3

InChI key

WOZZOSDBXABUFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Aluminum-tri-sec-butoxide is a metal organic chemical vapor deposition precursor and a chelating agent. It is widely used as a sol-gel precursor to deposit alumina thin films by atomic layer deposition method.

Application

Aluminum-tri-sec-butoxide can be used as a precursor :
  • To prepare Al2O3-based quasi-solid electrolyte for Li metal anodes. This ensures long cycle lifetime and high constant current density.
  • To synthesize nanospinel MgAl2O4 powder by a sol–gel method at low-temperature processing.
  • To fabricate transparent superhydrophobic aluminum oxide thin films to prevent soiling of solar panels.
  • To synthesize mesoporous alumina with large surface area to fabricate a functional interlayer for separator coating in a Li–S battery. It helps to suppress the shuttle effect and enhances cyclic stability andrateperformance.

Features and Benefits

  • Suitable for thin film deposition
  • Solubility in various solvents allows for the facile preparation of solution-based precursors
  • High thermal stability
  • Facilitates the anchoring of catalytic metal nanoparticles onto substrates

Has been utilized in making high-surface-area alumina using 2,4-pentanedione (Aldrich product P775-4) as a chelating agent.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chem. Abstr., 124, 328279f-328279f (1996)
Clark, H.W. et al.
Microporous Mater., 6, 115-115 (1996)
Polykarpos A Lazaridis et al.
Frontiers in chemistry, 6, 295-295 (2018-08-04)
The valorization of lignin that derives as by product in various biomass conversion processes has become a major research and technological objective. The potential of the production of valuable mono-aromatics (BTX and others) and (alkyl)phenols by catalytic fast pyrolysis of
Vaudry, F. et al.
Chemistry of Materials, 8, 1451-1451 (1996)
Zhao, G. Tohge, N.
Kobunshi Ronbunshu, 53, 253-253 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service