Skip to Content
Merck
All Photos(1)

Key Documents

202908

Sigma-Aldrich

Cadmium chloride

99.99% trace metals basis

Synonym(s):

Cadmium dichloride, Dichlorocadmium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CdCl2
Molecular Weight:
183.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid
solubility:
H2O: soluble 457 g/L at 20 °C (OECD Test Guideline 105)

grade

for analytical purposes

vapor density

6.3 (vs air)

vapor pressure

10 mmHg ( 656 °C)

Assay

99.99% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: cadmium

impurities

≤150.0 ppm Trace Metal Analysis

mp

568 °C (lit.)

solubility

H2O: soluble 457 g/L at 20 °C (OECD Test Guideline 105)

SMILES string

Cl[Cd]Cl

InChI

1S/Cd.2ClH/h;2*1H/q+2;;/p-2

InChI key

YKYOUMDCQGMQQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Cadmium chloride is colorless, odorless, and highly soluble in water. It is widely used in the field of catalysis and perovskite solar cells.

Application

Cadmium chloride can be used:
  • As an additive to fabricate perovskite solar cells to improve long-term stability and performance. The incorporation of Cd ions enhances optical absorption and carrier transfer.
  • As a precursor to synthesize functionalized cadmium telluride quantum dots for fluorescent immunosensors.
It can also be used as a catalyst for:
  • Aza-Michael reaction of electron-deficient alkene and a wide range of amines.
  • The one-pot synthesis of dihydropyrimidones via the Biginelli reaction.
  • The synthesis of benimidazoles.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cadmium chloride: An efficient catalyst for one-pot synthesis of 3,4-dihydropyrimidin-2 (1H)-ones
A Venkat Narsaiah, et al.
Synthesis, 2004, 1253-1256 (2004)
Bridget R Simon et al.
Toxicology reports, 1, 391-400 (2014-08-30)
Previously, we demonstrated the sensitivity of RPTEC/TERT1 cells, an immortalized human renal proximal tubule epithelial cell line, to two common environmental carcinogens, cadmium (Cd) and benzo[a]pyrene (B[a]P). Here, we measured BPDE-DNA adducts using a competitive ELISA method after cells were
Analysis of the decomposition gases from a and ?-Cd (BH 4) 2 synthesized by temperature controlled mechanical milling.
Didier Z, et al.
J. Alloy Compounds, 547, 76-80 (2013)
B R Simon et al.
Toxicology reports, 1, 231-242 (2014-08-16)
We have characterized initial canonical responses to two environmental toxicants, cadmium (Cd) and benzo[a]pyrene (B[a]P), in a novel
Introduction of cadmium chloride additive to improve the performance and stability of perovskite solar cells
Mustafa KA Mohammed, et al.
Royal Society of Chemistry Advances, 12, 20461-20470 (2022)

Articles

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service