Skip to Content
Merck
All Photos(1)

Key Documents

459852

Sigma-Aldrich

9,10-Anthracenedicarbonitrile

97%

Synonym(s):

9,10-Dicyanoanthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8N2
CAS Number:
Molecular Weight:
228.25
Beilstein:
1646384
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

340 °C (lit.)

SMILES string

N#Cc1c2ccccc2c(C#N)c3ccccc13

InChI

1S/C16H8N2/c17-9-15-11-5-1-2-6-12(11)16(10-18)14-8-4-3-7-13(14)15/h1-8H

InChI key

BIOPPFDHKHWJIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

9,10-Anthracenedicarbonitrile (DCA, ADC) is an anthracene derivative. Photochemical reactions of DCA in MeCN-MeOH or MeCN-H2O containing hydroxides or methoxides affords 9-methylimino-10-anthracenecarbonitrile. It participates in the generation of nucleophilic α-hydroxymethyl radicals from α-silyl ethers by irradiation. Binding energy of 9,10-anthracenedicarbonitrile adsorbed onto graphene is -1.23eV.

Application

9,10-Anthracenedicarbonitrile may be used as photosensitizer to investigate the photoisomerization of trans-stilbene (TS) in sodium dodecyl sulfate (SDS)/benzyl alcohol (BA)/H2O system.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

a-Silyl Ethers as Hydroxymethyl Anion Equivalents in Photoinduced Radical Electron Transfer Additions.
Gutenberger G, et al.
Angewandte Chemie (International Edition in English), 37(5), 660-662 (1998)
Xia Guo et al.
Journal of colloid and interface science, 283(2), 578-584 (2005-02-22)
Photoisomerization of trans-stilbene (TS) was investigated in sodium dodecyl sulfate (SDS)/benzyl alcohol (BA)/H(2)O systems in order to establish the relationship between the reaction yields and the compositions and structures of molecular organized assemblies. The results show that, in SDS/BA/H(2)O systems
Photoinduced Bimolecular Electron Transfer from Cyano Anions in Ionic Liquids.
Boning W, et al.
The Journal of Physical Chemistry B, 119, 14790-14799 (2015)
Steven Bailey et al.
The Journal of chemical physics, 140(5), 054708-054708 (2014-02-12)
To identify families of stable planar anchor groups for use in single molecule electronics, we report detailed results for the binding energies of two families of anthracene and pyrene derivatives adsorbed onto graphene. We find that all the selected derivatives
The photochemical reactions of 9, 10-anthracenedicarbonitrile and 1, 4-naphthalenedicarbonitrile in acetonitrile in the presence of bases.
Freccero M, et al.
Tetrahedron, 50(7), 2115-2130 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service