Skip to Content
Merck
All Photos(3)

Key Documents

903167

Sigma-Aldrich

10-Phenylphenothiazine

≥95%

Synonym(s):

10-Phenyl-10H-phenothiazine, N-Phenylphenothiazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13NS
CAS Number:
Molecular Weight:
275.37
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Photocatalysis

mp

94 °C

photocatalyst activation

365 nm

SMILES string

C12=CC=CC=C1SC3=C(C=CC=C3)N2C4=CC=CC=C4

InChI

1S/C18H13NS/c1-2-8-14(9-3-1)19-15-10-4-6-12-17(15)20-18-13-7-5-11-16(18)19/h1-13H

InChI key

WSEFYHOJDVVORU-UHFFFAOYSA-N

Application

An organic photo-catalyst used in synthesis of small molecules as well as polymerization reactions.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Honghong Gong et al.
Angewandte Chemie (International ed. in English), 57(1), 333-337 (2017-11-15)
Fluorinated polymers are important materials that are widely used in many areas. Herein, we report the development of a metal-free photocontrolled radical polymerization of semifluorinated (meth)acrylates with a new visible-light-absorbing organocatalyst. This method enabled the production of a variety of
Single-Ion Homopolymer Electrolytes with High Transference Number Prepared by Click Chemistry and Photoinduced Metal-Free Atom-Transfer Radical Polymerization.
Li. S, et al.
ACS Energy Letters, 3(1), 20-27 (2018)
Hengbin Wang et al.
Journal of the American Chemical Society, 140(1), 163-166 (2017-12-20)
We describe a new catalytic approach to selective functionalization of the strong C-F bonds in trifluoromethylaromatic (Ar-CF3) systems. In this approach, single electron reduction of Ar-CF3 substrates (using a photoredox catalyst) results in difluorobenzylic radical formation through a C-F cleavage
Mao Chen et al.
ACS central science, 3(2), 124-134 (2017-03-11)
Light-initiated additive manufacturing techniques typically rely on layer-by-layer addition or continuous extraction of polymers formed via nonliving, free radical polymerization methods that render the final materials "dead" toward further monomer insertion; the polymer chains within the materials cannot be reactivated
Organocatalyzed Photoredox Polymerization from Aromatic Sulfonyl Halides: Facilitating Graft from Aromatic C-H bonds.
Zhao Y, et al.
Macromolecules, 51(3), 938-946 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service