Skip to Content
Merck
All Photos(3)

Key Documents

391980

Sigma-Aldrich

1,2,4,5-Tetracyanobenzene

97%

Synonym(s):

1,2,4,5-Benzenetetracarbonitrile, Pyromellitic acid tetranitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H2(CN)4
CAS Number:
Molecular Weight:
178.15
Beilstein:
1875027
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

265-268 °C (lit.)

solubility

acetone: soluble 25 mg/mL, clear, colorless to yellow

functional group

nitrile

SMILES string

N#Cc1cc(C#N)c(cc1C#N)C#N

InChI

1S/C10H2N4/c11-3-7-1-8(4-12)10(6-14)2-9(7)5-13/h1-2H

InChI key

FAAXSAZENACQBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2,4,5-Tetracyanobenzene (TCNB, TCB) is a tetra substituted benzene. The IR and raman spectra of TCNB and its 1:1 complex with durene has been recorded. The time-resolved linear dichroism spectroscopic study of hexamethylbenzene-1,2,4,5-tetracyanobenzene charge-transfer complex has been investigated. The influence of magnetic field on the charge-transfer fluorescence of 1,2,4,5-tetracyanobenzene-doped poly(N-vinylcarbazole) thin film has been studied.

Application

1,2,4,5-Tetracyanobenzene (TCB, TCNB) is suitable reagent used in the synthesis of o-3,4-dimethyltetrathiafulvalene (o-Me2TTF)-1,2,4,5-tetracyanobenzene (TCNB) 1:1 complex and radiative exciplexes of TCB in non-polar solvents.

1,2,4,5-Tetracyanobenzene (Pyromellitic acid tetranitrile, PMTN, TCB, TCNB, 1,2,4,5-benzenetetracarbonitrile ) may be used in the following studies:
  • Solid-state diffusion of TCNB into 9-methylanthracene molecular crystal nanorods forming cocrystal nanorods.
  • Synthesis of copper polyphthalocyanine (CuPPC) thin films, an elementoorganic semiconductor.
  • Preparation of charge transfer (CT) nanocrystallites which may have a potential use in nanofluidics to observe the rotational motion of nanoobjects.
  • Hydrothermal synthesis of Co2(terpy)2(btec)•H2O (terpy = 2,2′:6′,2"-terpyridine, btec = 1,2,4,5-benzenetetracarboxylate).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inelastic and quasielastic neutron scattering and IR and R spectroscopic studies of 1, 2, 4, 5-tetracyanobenzene (TCNB)-1, 2, 4, 5-tetramethylbenzene (durene) complex.
Bator G, et al.
Phase Transitions, 80(6-7), 489-500 (2007)
Structural Studies of the 1: 1 Complex of o-3, 4-Dimethyltetrathiafulvalene (o-Me2TTF) and 1, 2, 4, 5-Tetracyanobenzene (TCNB).
Reinheimer EW, et al.
Journal of Chemical Crystallography, 40(6), 514-519 (2010)
Formation of cocrystal nanorods by solid-state reaction of tetracyanobenzene in 9-methylanthracene molecular crystal nanorods.
Al-Kaysi RO, et al.
Crystal Growth & Design, 9(4), 1780-1785 (2009)
Fuyuki Ito et al.
Journal of the American Chemical Society, 125(16), 4722-4723 (2003-04-17)
Magnetic field effects on the charge-transfer (CT) fluorescence of a 1,2,4,5-tetracyanobenzene-doped poly(N-vinylcarbazole) thin film were investigated to clarify the primary process in photoconductive organic amorphous solid. The CT fluorescence increased with increasing magnetic field until 10 mT, and then it
Synthesis and structure of high-quality films of copper polyphthalocyanine-2D conductive polymer.
Sedlovets DM, et al.
Materials Research Bulletin, 48(10), 3955-3960 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service