Skip to Content
Merck
All Photos(2)

Key Documents

724440

Sigma-Aldrich

9,10-Anthracenedicarboxylic acid

greener alternative

95%

Synonym(s):

H2ADC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10O4
CAS Number:
Molecular Weight:
266.25
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>300 °C

greener alternative category

SMILES string

OC(=O)c1c2ccccc2c(C(O)=O)c3ccccc13

InChI

1S/C16H10O4/c17-15(18)13-9-5-1-2-6-10(9)14(16(19)20)12-8-4-3-7-11(12)13/h1-8H,(H,17,18)(H,19,20)

InChI key

FDFGHPKPHFUHBP-UHFFFAOYSA-N

General description

9,10-Anthracenedicarboxylic acid (H2L) is an anthracene based dicarboxylic compound, which has a larger conjugating π-system that enables the development of fluorescent materials. It has interesting magnetic and luminescent properties. It can be used as a bridging carboxylic acid ligand with a steric bulk due to the presence of its anthracene ring.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

H2L can be potentially used in the formation of highly porous nanotubular and ultramicroporous metal organic frameworks (MOFs) for greener applications like hydrogen storage, methane storage and gas separation.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultramicroporous Metal- Organic Framework Based on 9, 10-Anthracenedicarboxylate for Selective Gas Adsorption
Ma S, et al.
Inorganic Chemistry, 46(21), 8499-8501 (2007)
Manganese (II) complexes with a bulky anthracene-based dicarboxylic ligand: Syntheses, crystal structures, and magnetic properties
Liu C, et al.
Australian Journal of Chemistry, 61(5), 382-390 (2008)
Cadmium (II) and lanthanum (III) coordination architectures with anthracene-9, 10-dicarboxylate: Crystal structures and photoluminescent properties
Wang J, et al.
Inorgorganica Chimica Acta, 385(6), 58-64 (2012)
A nanotubular metal-organic framework with permanent porosity: structure analysis and gas sorption studies
Ma S, et al.
Chemical Communications (Cambridge, England), 385(27), 4049-4051 (2009)
Zinc (II) coordination architectures with two bulky anthracene-based carboxylic ligands: crystal structures and luminescent properties
Wang J, et al.
CrystEngComm, 10(6), 681-692 (2008)

Articles

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Metal-organic frameworks offer high surface area materials for alternative energy applications through simple synthetic strategies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service