Skip to Content
Merck
All Photos(1)

Key Documents

456845

Sigma-Aldrich

Zinc chloride

AnhydroBeads, amorphous, −10 mesh, 99.999% trace metals basis

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
amorphous
solubility:
H2O: soluble
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 428 °C)

Quality Level

product line

AnhydroBeads

Assay

99.999% trace metals basis

form

amorphous

reaction suitability

reagent type: catalyst
core: zinc

impurities

≤15.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

293 °C (lit.)

solubility

H2O: soluble

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Davies et al.
Cell death & disease, 2, e136-e136 (2011-04-01)
The activation and regulation of target genes by the tumour-suppressor p53 dictates the fate of a cell, with cell cycle arrest or apoptosis being two distinct outcomes. PERP (p53 apoptosis effector related to PMP-22), a p53 transcriptional target, is induced
Michele K Sankako et al.
Pharmacological reports : PR, 64(6), 1537-1546 (2013-02-15)
The aim of this study was to evaluate the changes in testicular function of rats due to cigarette smoke exposure and the possible mechanism by which zinc protects against these alterations. Male Wistar rats (60 days old) were randomly divided
Hai-Long Jiang et al.
Journal of the American Chemical Society, 134(36), 14690-14693 (2012-08-22)
Reactions of ZrCl(4) and single or mixed linear dicarboxylic acids bearing methyl or azide groups lead to highly stable isoreticular metal-organic frameworks (MOFs) with content-tunable, accessible, reactive azide groups inside the large pores. These Zr-based MOFs offer an ideal platform
Kaushik Nath et al.
Environmental science and pollution research international, 20(6), 4030-4045 (2012-12-06)
Utilization of agrowaste materials for the production of activated carbon, as an excellent adsorbent with large surface area, is well established industrially, for dephenolation of wastewater. In the present work, dried pods of Prosopis cineraria-a novel and low-cost agrowaste material-were
Jessica Duprez et al.
PloS one, 7(10), e46831-e46831 (2012-10-12)
Rat pancreatic islet cell apoptosis is minimal after prolonged culture in 10 mmol/l glucose (G10), largely increased in 5 mmol/l glucose (G5) and moderately increased in 30 mmol/l glucose (G30). This glucose-dependent asymmetric V-shaped profile is preceded by parallel changes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service