Skip to Content
Merck
All Photos(1)

Key Documents

429430

Sigma-Aldrich

Zinc chloride

anhydrous, powder, ≥99.995% trace metals basis

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.995% trace metals basis
grade:
anhydrous
form:
powder
solubility:
acetone: slightly soluble(lit.)
ethanol: slightly soluble(lit.)

grade

anhydrous

Quality Level

vapor pressure

1 mmHg ( 428 °C)

Assay

≥99.995% trace metals basis

form

powder

impurities

≤50.0 ppm Trace Metal Analysis

bp

732 °C/1 atm (lit.)

mp

293 °C (lit.)

solubility

acetone: slightly soluble(lit.)
ethanol: slightly soluble(lit.)

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Electrodeposition of zinc on glassy carbon and nickel substrates in zinc chloride-1-ethyl-3-methylimidazolium chloride molten salt is studied.

Application

Zinc Chloride may be used:
  • as catalyst in knoevenagel condensation of carbonyl substrates with acidic methylene reagents
  • in the preparation of porous carbon nanofibers, useful in the fabrication of efficient electrodes for supercapacitors
  • as a catalyst in preparation of poly(propylene fumarate)
  • in the low temperature synthesis of nanocrystalline zinc oxide films
  • in the synthesis of zinc oxide nanoparticles with low agglomeration. Aqueous suspensions of the nanoparticles displayed high transmittance in the visible light range, but exhibited strong absorption in the UV range.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tsuzuki, T., et al.
J. Autralasian Ceram. Soc., 38, 15-15 (2002)
Gorbik, P.P., et al.
Dopovidi Natsional'noi Akademii Nauk Ukranaie, 1, 135-135 (2004)
Electrodeposition of zinc from a Lewis acidic zinc chloride-1-ethyl-3-methylimidazolium chloride molten salt.
Lin Y-F and Sun I.
Electrochimica Acta, 44(16), 2771-2777 (1999)
Self-Sustained Thin Webs Consisting of Porous Carbon Nanofibers for Supercapacitors via the Electrospinning of Polyacrylonitrile Solutions Containing Zinc
Kim C, et al.
Advanced Mat., 19(17), 2341-2346 (2007)
Zinc chloride as a new catalyst for Knoevenagel condensation.
Shanthan RP and Venkataratnam RV.
Tetrahedron Letters, 32(41), 5821-5822 (1991)

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service