Skip to Content
Merck
All Photos(1)

Documents

230022

Sigma-Aldrich

Zinc bromide

99.999% trace metals basis

Synonym(s):

Zinc dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnBr2
CAS Number:
Molecular Weight:
225.20
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38150410
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: zinc

impurities

≤15.0 ppm Trace Metal Analysis

bp

~670 °C/1 atm (lit.)

mp

394 °C (lit.)

solubility

diethyl ether: slightly soluble(lit.)
ethanol: very soluble(lit.)

SMILES string

Br[Zn]Br

InChI

1S/2BrH.Zn/h2*1H;/q;;+2/p-2

InChI key

VNDYJBBGRKZCSX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Zinc bromide supported on silica or montmorillonite can be used as a selective catalyst for the bromination of aromatic substrates.

It can be used as starting material to prepare poly(ethylene oxide)/ zinc salt systems, which can be used as polymer electrolytes.

Itcan also be used to prepare ZnO nanoparticles.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franz, A.K. Woerpel, K.A.
Journal of the American Chemical Society, 121, 949-949 (1999)
Frederic G Buono et al.
Organic letters, 10(23), 5325-5328 (2008-11-05)
Kinetic studies using reaction calorimetry were conducted under synthetically relevant conditions to study the effect of additives in the cyanation of bromobenzene catalyzed by palladium complexes. This work demonstrates that the addition of a catalytic amount of ZnBr(2) facilitates the
Direct synthesis of alkynylstannanes: ZnBr2 catalyst for the reaction of tributyltin methoxide and terminal alkynes.
Kensuke Kiyokawa et al.
Angewandte Chemie (International ed. in English), 50(44), 10393-10396 (2011-09-15)
Isabelle Aillaud et al.
Molecules (Basel, Switzerland), 15(11), 8144-8155 (2010-11-13)
A methodological study devoted to the Mannich-like multicomponent synthesis of the antiplatelet agent (±)‑clopidogrel (7) and the ethyl ester analogue 6 is described. The process involves the formation of 2-chlorophenyl zinc bromide (2) and its subsequent reaction with an alkyl
Toshiaki Sasada et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(34), 9385-9394 (2011-07-23)
The [5+1] annulation of enamidines, which were prepared from functionalized silanes, organolithium compounds and two nitriles, with N,N-dimethylformamide dialkyl acetals as the C1 unit is described, leading to the synthesis of tri- and tetrasubstituted pyrimidine derivatives under catalyst- and solvent-free

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service