This product is a 0.1 M solution prepared in water. No additional components have been added.
39059
Zinc chloride 0.1 M solution
Synonym(s):
Zinc chloride solution, Additive Screening Solution 29/Kit-No 78374
About This Item
Recommended Products
form
liquid
Quality Level
density
1.01 g/mL at 20 °C
storage temp.
2-8°C
SMILES string
Cl[Zn]Cl
InChI
1S/2ClH.Zn/h2*1H;/q;;+2/p-2
InChI key
JIAARYAFYJHUJI-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Application
- Zinc, sulfur and nitrogen co-doped carbon from sodium chloride/zinc chloride-assisted pyrolysis of thiourea/sucrose for highly efficient oxygen reduction reaction in both acidic and alkaline media.: This study details the use of zinc chloride in a 0.1 M solution to assist in the pyrolysis process for creating co-doped carbon materials, which show high efficiency in oxygen reduction reactions. This has significant implications for fuel cell technologies and other electrochemical applications (Cao et al., 2020).
- The affinity of histamine for Gq protein-coupled histamine H(1)-receptors is predominantly regulated by their internalization in human astrocytoma cells.: This research explores the role of zinc chloride 0.1 M solution in modulating histamine receptor activity in astrocytoma cells, providing insights into cellular signaling mechanisms and potential therapeutic targets (Hishinuma et al., 2012).
- Effect of calcium chloride, zinc chloride, and water infusion on metmyoglobin reducing activity and fresh lamb color.: The paper investigates the impact of a 0.1 M zinc chloride solution on meat color and quality, demonstrating its potential to enhance metmyoglobin reducing activity and improve the visual appeal of fresh lamb (Bekhit et al., 2005).
- Furin inhibition by compounds of copper and zinc.: This study shows how a 0.1 M solution of zinc chloride can inhibit the activity of furin, a protease involved in various biological processes, which has implications for developing therapeutic inhibitors for diseases involving protease activity (Podsiadlo et al., 2004).
- Release of hyaluronidase during in vitro development of Ascaris suum from the third to fourth larval stage.: Research on the in vitro development of Ascaris suum larvae highlights the role of zinc chloride 0.1 M solution in facilitating the release of hyaluronidase, an enzyme critical for larval development, shedding light on potential parasitic intervention strategies (Rhoads et al., 2001).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the ZnCl2 in? Water, or some organic solvent? Thanks.
1 answer-
Helpful?
-
-
What is this dissolve in?
1 answer-
This product is a 0.1 M solution in water.
Helpful?
-
-
Is this product protease free?
1 answer-
This product is not tested for protease activity. Should this be a requirement, see the link below to review product Z0152, the protease-free powder form:
https://www.sigmaaldrich.com/product/sigma/z0152Helpful?
-
-
こちらの商品の溶媒は何になりますでしょうか?
1 answer-
This product is a 0.1 M solution in water.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service