Skip to Content
Merck
All Photos(2)

Key Documents

203149

Sigma-Aldrich

Copper(II) chloride

99.999% trace metals basis

Synonym(s):

Cupric chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl2
CAS Number:
Molecular Weight:
134.45
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38150111
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: copper

impurities

≤15.0 ppm Trace Metal Analysis

mp

620 °C (lit.)

density

3.386 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[Cu]Cl

InChI

1S/2ClH.Cu/h2*1H;/q;;+2/p-2

InChI key

ORTQZVOHEJQUHG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Copper chloride is a white solid. It is insoluble in water. Copper(II) chloride may be prepared by dissolving excess of copper (II) oxide or copper(II) carbonate in dilute hydrochloric acid.

Application

Adding CuCl2 to AgNO3 with EG1mYAbe is used to produce uniform Ag nanowires. Copper chloride may be used as a catalyst in organic chlorination reactions. Used with palladium in a catalytic synthesis of 3-haloindoles via an annulation process.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noble-Metal Nanostructures with Controlled Morphologies
Jiang M, et al.
Material Matters, 4(1) null
Handbook of Copper Compounds and Applications null
Chemistry (Weinheim An Der Bergstrasse, Germany) null
Synthesis, 1841-1841 (2007)
Iron(II)-catalyzed trifluoromethylation of potassium vinyltrifluoroborates.
Andrew T Parsons et al.
Angewandte Chemie (International ed. in English), 51(12), 2947-2950 (2012-02-14)

Articles

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

See All

Related Content

This application note discusses the synthesis of silver nanowires.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service