Skip to Content
Merck
All Photos(3)

Key Documents

696668

Sigma-Aldrich

Nickel(II) chloride ethylene glycol dimethyl ether complex

98%

Synonym(s):

Dichloro(dimethoxyethane)nickel, NiCl2 glyme

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiCl2:CH3OCH2CH2OCH3
CAS Number:
Molecular Weight:
219.72
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

reaction suitability

core: nickel
reagent type: catalyst

mp

>300 °C

SMILES string

Cl[Ni]Cl.COCCOC

InChI

1S/C4H10O2.2ClH.Ni/c1-5-3-4-6-2;;;/h3-4H2,1-2H3;2*1H;/q;;;+2/p-2

InChI key

OCMNCWNTDDVHFK-UHFFFAOYSA-L

Application

Nickel(II) chloride ethylene glycol dimethyl ether complex can be used:
  • As a catalyst for the borylation of racemic benzylic chloride to synthesize enantioenriched benzylic boronic esters.
  • As a promoter for the trifluoromethylation of alkyl iodides to synthesize broad range of alkyl-CF3 compounds.
  • For the synthesis of nickel bis(benzimidazol-2-ylidene) pincer complexes which can be used for electrocatalytic reduction of CO2 to CO.
  • As a Lewis acid catalyst for C-acylation β-ketoesters through photoactivation by visible light.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Carc. 1B - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and comparison of nickel, palladium, and platinum bis (N-heterocyclic carbene) pincer complexes for electrocatalytic CO 2 reduction.
Therrien JA, et al.
Dalton Transactions, 47(6), 1827-1840 (2018)
Amanda Grass et al.
Dalton transactions (Cambridge, England : 2003), 50(7), 2501-2509 (2021-01-31)
Reaction of LiOCtBu2Ph with TlPF6 forms the dimeric Tl2(OCtBu2Ph)2 complex, a rare example of a homoleptic thallium alkoxide complex demonstrating formally two-coordinate metal centers. Characterization of Tl2(OCtBu2Ph)2 by 1H and 13C NMR spectroscopy and X-ray crystallography reveals the presence of
Catalyst-Controlled Regioselective Acylation of ?-Ketoesters with ?-Diazo Ketones Induced by Visible Light.
Liu D, et al.
Organic Letters, 20(22), 7278-7282 (2018)
Nickel-Catalyzed Enantioconvergent Borylation of Racemic Secondary Benzylic Electrophiles.
Wang Z, et al.
Angewandte Chemie (International Edition in English), 57(44), 14529-14532 (2018)
Copper-Catalyzed Reductive Trifluoromethylation of Alkyl Iodides with Togni?s Reagent.
Chen Y, et al.
Organic Letters, 20(15), 4677-4680 (2018)

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service