Skip to Content
Merck
All Photos(3)

Documents

44242

Sigma-Aldrich

Dodecyltrimethylammonium chloride

≥99.0% (AT)

Synonym(s):

DTAC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11N(CH3)3Cl
CAS Number:
Molecular Weight:
263.89
Beilstein:
3915951
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

description

cationic

Assay

≥99.0% (AT)

form

solid

mol wt

263.89 g/mol

mp

37 °C

solubility

H2O: 50 mg/mL, clear, colorless

SMILES string

[Cl-].CCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C15H34N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;/h5-15H2,1-4H3;1H/q+1;/p-1

InChI key

DDXLVDQZPFLQMZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Dodecyltrimethylammonium chloride is a cationic alkyltrimethylammonium chloride surfactant that can undergo micellization in aqueous solution. it is mainly used for the denaturation of proteins.

Application

Dodecyltrimethylammonium chloride has also been used as a cationic surfactant in:
  • Transfer hydrogenation reactions of ketones/aldehydes.
  • Ring-opening metathesis polymerization of olefins.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aqueous olefin metathesis
Burtscher D and Grela K
Angewandte Chemie (International Edition in English), 48, 442-454 (2009)
Effect of temperature on critical micelle concentration and thermodynamic behavior of dodecyldimethylethylammonium bromide and dodecyltrimethylammonium chloride in aqueous media.
Mehta S
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 255(1-3), 153-157 (2005)
Temperature and salt-induced micellization of dodecyltrimethylammonium chloride in aqueous solution: A thermodynamic study.
Sarac B
Journal of Colloid and Interface Science, 338(1), 216-221 (2009)
Phase separation in the isolation and purification of membrane proteins.
Arnold T
Biotechniques, 43(4), 427-442 (2007)
Transfer-hydrogenation reactions of ketones/aldehydes in water using first generation ruthenium indenylidene olefin metathesis catalyst: one step towards sequential cross-metathesis/transfer hydrogenation reactions
Ozturk B and Ozturk S
Molecular Catalysis, 480, 110640-110640 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service