Skip to Content
Merck
All Photos(3)

Key Documents

13380

Sigma-Aldrich

Benzyldimethyldodecylammonium chloride

≥99.0% (AT)

Synonym(s):

Benzododecinium chloride, Benzyldodecyldimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11N(Cl)(CH3)2CH2C6H5
CAS Number:
Molecular Weight:
339.99
Beilstein:
3921689
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

description

cationic

Assay

≥99.0% (AT)

form

solid

mol wt

339.99 g/mol

SMILES string

[Cl-].CCCCCCCCCCCC[N+](C)(C)Cc1ccccc1

InChI

1S/C21H38N.ClH/c1-4-5-6-7-8-9-10-11-12-16-19-22(2,3)20-21-17-14-13-15-18-21;/h13-15,17-18H,4-12,16,19-20H2,1-3H3;1H/q+1;/p-1

InChI key

JBIROUFYLSSYDX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Benzyldimethyldodecylammonium chloride, a quaternary ammonium compound (QAC), is a cationic surfactant. It can be used as a:

  • corrosion inhibitor.
  • model for benzyltrialkylammonium salts to study the thermal degradation pathway of benzyl QACs.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Nemes et al.
Die Pharmazie, 73(1), 16-18 (2018-02-15)
Aqueous pharmaceutical solutions provide prosperous living conditions for microbiological agents. In order to eliminate these microbes, we use preservatives which can harm human cells as well. Their cytotoxicity is concentration-dependent and the aim of our study was to find how
Thermal degradation of some benzyltrialkylammonium salts using pyrolysis-gas chromatography-mass spectrometry.
Haskins NJ and Mitchell R
Analyst, 116(9), 901-903 (1991)
Quaternary ammonium cationic surfactants increase bioactivity of indoxacarb on pests and toxicological risk to Daphnia magna.
Li B, et al.
Ecotoxicology and Environmental Safety, 149(5), 190-196 (2018)
Adsorption of benzyldimethyldodecylammonium chloride onto stainless steel using the quartz crystal microbalance and the depletion methods: an optimisation study.
Salama IE, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 447(5), 155-165 (2014)
Anton G Rogov et al.
Oxidative medicine and cellular longevity, 2020, 8956504-8956504 (2020-02-28)
Benzalkonium chloride (BAC) is currently the most commonly used antimicrobial preservative in ophthalmic solutions, nasal sprays, and cosmetics. However, a large number of clinical and experimental investigations showed that the topical administration of BAC-containing eye drops could cause a variety

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service