Skip to Content
Merck
All Photos(1)

Key Documents

30725

Sigma-Aldrich

Decyltrimethylammonium bromide

≥98.0% (NT)

Synonym(s):

N,N,N-Trimethyl-1-decanaminium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9N(CH3)3(Br)
CAS Number:
Molecular Weight:
280.29
Beilstein:
3915222
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (NT)

SMILES string

[Br-].CCCCCCCCCC[N+](C)(C)C

InChI

1S/C13H30N.BrH/c1-5-6-7-8-9-10-11-12-13-14(2,3)4;/h5-13H2,1-4H3;1H/q+1;/p-1

InChI key

PLMFYJJFUUUCRZ-UHFFFAOYSA-M

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mandy H M Leung et al.
Langmuir : the ACS journal of surfaces and colloids, 24(11), 5672-5675 (2008-05-08)
The alkaline hydrolysis of curcumin was studied in three types of micelles composed of the cationic surfactants cetyl trimethylammonium bromide (CTAB) and dodecyl trimethylammonium bromide (DTAB) and the anionic surfactant sodium dodecyl sulfate (SDS). At pH 13, curcumin undergoes rapid
Kabir-ud-Din et al.
Colloids and surfaces. B, Biointerfaces, 92, 16-24 (2011-12-14)
The micellization behaviors of two amphiphilic drugs ((amitriptyline hydrochloride (AMT) and imipramine hydrochloride (IMP)) in presence of cationic surfactants (conventional as well as gemini) have been investigated conductometrically at four mole fractions and four temperatures. The critical micelle concentration (cmc)
Ting Zhou et al.
Journal of colloid and interface science, 389(1), 175-181 (2012-10-10)
The interactions of BSA with a series of cationic imidazolium gemini surfactants ([C(n)-s-C(n)im]Br(2), n=10, 12, 14, s=2, 4, 6), quaternary ammonium surfactants (C(12)C(2)C(12)), and their corresponding monomers ([C(12)mim]Br and DTAB) are investigated by fluorescence using pyrene as a molecular probe
Sergii Rudiuk et al.
Journal of colloid and interface science, 368(1), 372-377 (2011-11-11)
We study the compaction of genomic DNA by a series of alkyltrimethylammonium bromide surfactants having different hydrocarbon chain lengths n: dodecyl-(DTAB, n=12), tetradecyl-(TTAB, n=14) and hexadecyl-(CTAB, n=16), in the absence and in the presence of negatively charged silica nanoparticles (NPs)
Parvaiz Ahmad Bhat et al.
The journal of physical chemistry. B, 113(4), 997-1006 (2009-01-07)
Mixed surfactants may improve the performance of surfactant-enhanced solubilization of drugs and thus can serve as the tool for increased bioavalaibility, controlled drug release, and targeted delivery. Solubilization of Naproxen by micellar solutions at 25 degrees C using single and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service