Skip to Content
Merck
All Photos(1)

Key Documents

86694

Sigma-Aldrich

Tetraethylene glycol monododecyl ether

BioXtra, ≥98.0% (GC)

Synonym(s):

C12E4, Dodecyl tetraethylene glycol ether, Dodecyltetraglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11(OCH2CH2)4OH
CAS Number:
Molecular Weight:
362.54
Beilstein:
1791369
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

product line

BioXtra

Assay

≥98.0% (GC)

form

liquid

mol wt

362.54 g/mol

refractive index

n20/D 1.450-1.454

density

0.946 g/mL at 20 °C (lit.)

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCO

InChI

1S/C20H42O5/c1-2-3-4-5-6-7-8-9-10-11-13-22-15-17-24-19-20-25-18-16-23-14-12-21/h21H,2-20H2,1H3

InChI key

WPMWEFXCIYCJSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetraethylene glycol monododecyl ether (C12E4) is a nonionic surfactant. It is used as a stabilizer in several phase behavior and evaporation studies. This polyoxyethylated lauryl ether surfactant is a water-soluble surfactant.

Application

Tetraethylene glycol monododecyl ether (C12E4) has been used:


  • as a water-soluble surfactant in computational and physical experiments to report the post-deposition motion of the surfactant front and the deformation of the subphase surface using Marangoni flow and capillary waves
  • to study its mixed micellization and interfacial properties in the absence and presence of sodium propionate
  • to investigate the phase transformation of C12E4 aqueous solutions by Rayleigh light scattering (RLS) when adopting different heating ways

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Sawayama et al.
Langmuir : the ACS journal of surfaces and colloids, 27(14), 8653-8658 (2011-06-09)
By mixing a small volume of THF containing guanosine derivative 1 and tetraethylenegrycol dodecyl ether (TEGDE) with water and subsequently removing TEGDE by gel permeation chromatography, micrometer-sized giant unilamellar vesicles (GUV) of 1 were successfully prepared. The vesicle membrane was
N Taulier et al.
Biophysical journal, 78(2), 857-865 (2000-02-02)
We have used a lamellar phase made of a nonionic surfactant, dodecane and water, as a model membrane to investigate its interactions with macromolecular inclusions bringing together two membranes, i.e., acting as macromolecular snaps. In systems devoid of inclusions, the
Aijaz A Dar et al.
Journal of colloid and interface science, 298(1), 395-405 (2005-12-28)
Mixed micelle formation and interfacial properties of aqueous binary surfactant combinations of dodecyltrimethylammonium bromide (C12TAB) and tetraethyleneglycol mono-n-dodecyl ether (C12E4) at 30 degrees C in absence and presence of sodium propionate (NaPr) have been investigated. The critical micelle concentration, aggregation
Phase transformation of tetraethyleneglycol dodecyl ether solution studied by light scattering spectra: Micelle aggregation, vesicle and lamellar phase
Zhou J, et al.
Journal of Molecular Structure, 987, 91-100 (2011)
Caroline Senac et al.
Langmuir : the ACS journal of surfaces and colloids, 33(39), 10225-10238 (2017-08-24)
Polyoxyethylene glycol alkyl ether amphiphiles (C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service