Skip to Content
Merck
All Photos(5)

Documents

440744

Sigma-Aldrich

Polycaprolactone

average Mn 80,000

Synonym(s):

2-Oxepanone homopolymer, 6-Caprolactone polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O2)n
UNSPSC Code:
12162002
NACRES:
NA.23

form

pellets (~3 mm)

mol wt

average Mn 80,000

impurities

<0.5% water

mp

60 °C (lit.)

density

1.145 g/mL at 25 °C

Mw/Mn

<2

application(s)

3D bioprinting
advanced drug delivery

SMILES string

C1CCC(=O)OCC1 (Canonical-SMILES)

InChI

1S/C6H10O2/c7-6-4-2-1-3-5-8-6/h1-5H2

InChI key

PAPBSGBWRJIAAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polycaprolactone (PCL) is a semi crystalline hydrophobic biodegradable polyester which finds major applications as a drug delivery agent because of its cost efficiency, high toughness and biocompatibility. PCL possesses some unique properties such as:
  • It degrades slower than other biodegradable polyesters in physiological condition, this property can be exploited in the controlled release of drugs in target tissues over a period of time.
  • Its poor surface wetting and interaction with biological fluids on account of its hydrophobicity leads to poor cell adhesion and proliferation, hence it is blended with other synthetic /natural polymers.

Application

  • PCL loaded with antibiotics may be used to treat infections of the respiratory tract, like tuberculosis.
  • Investigations were carried out based upon phenotypic responses of human bone marrow mesenchymal cells (hBMSCs) for different ratio of chitosan/ polycaprolactone (PCL) blends.
  • PCL/biomedical ceramic materials have been studied for possible osteo tissue regeneration.
  • Action of PCL/graded insulin/beta-5 glycerophosphate concentrations on osteochondral tissue formation through adipose-derived stromal cell differentiation.
  • Other general uses include:extrusion aid, die lubricant, mold release, pigment and filler dispersion aid and polyester segments in urethanes and block polyesters.

Features and Benefits

Biodegradable polymer
Non-toxic, biodegradable in soil, broad miscibility, mechanical compatibility with many polymers and good adhesion to a broad spectrum of substrates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hung-Jen Shao et al.
Journal of biomedical materials research. Part A, 100(12), 3344-3352 (2012-06-27)
The phenotypic responses of human bone marrow mesenchymal cells (hBMSCs) on different ratio of chitosan/polycaprolactone (PCL) blends were investigated in this study. The results showed that hBMSCs existed different morphology on chitosan/PCL blends due to the different adhesion characteristic of
Cevat Erisken et al.
Tissue engineering. Part A, 17(9-10), 1239-1252 (2010-12-30)
The ability to fabricate tissue engineering scaffolds containing systematic gradients in the distributions of stimulators provides additional means for the mimicking of the important gradients observed in native tissues. Here the concentration distributions of two bioactive agents were varied concomitantly
Muhammad Shafiq et al.
Arteriosclerosis, thrombosis, and vascular biology, 38(7), e117-e134 (2018-06-02)
The objective of this study was to develop small-diameter vascular grafts capable of eluting SDF (stromal cell-derived factor)-1α-derived peptide and SP (substance P) for in situ vascular regeneration. Polycaprolactone (PCL)/collagen grafts containing SP or SDF-1α-derived peptide were fabricated by electrospinning.
T Limongi et al.
Molecular neurobiology, 55(12), 8788-8798 (2018-03-31)
Biopolymers are increasingly employed for neuroscience applications as scaffolds to drive and promote neural regrowth, thanks to their ability to mediate the upload and subsequent release of active molecules and drugs. Synthetic degradable polymers are characterized by different responses ranging
Jixin Xue et al.
Biomaterials, 34(11), 2624-2631 (2013-01-29)
Tissue engineering approach continuously requires for emerging strategies to improve the efficacy in repairing and regeneration of tissue defects. Previously, we developed a sandwich model strategy for cartilage engineering, using the combination of acellular cartilage sheets (ACSs) and chondrocytes. However

Articles

In the past two decades, tissue engineering and regenerative medicine have become important interdisciplinary fields that span biology, chemistry, engineering, and medicine.

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service